Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu480993_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-21a-3p
          Stem-loop Accession Number: MI0000569
          miRBase Version: v22.1
          Chromosome Location: Chr.11: 86584067 - 86584158 [-] on Build GRCm38
          Mature miRNA Sequence: CAACAGCAGUCGAUGGGCUGUC
          Species: Mouse, Rat, Artibeus jamaicensis, Cricetulus griseus, Dasypus novemcinctus, Eptesicus fuscus, Guinea pig, Ornithorhynchus anatinus, Rabbit
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu480993_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000060, mir-21

          Mature miRNA Sequence:

          CAACAGCAGUCGAUGGGCUGUC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-21a-3p MIMAT0004628
          Rat rno-miR-21-3p MIMAT0004711
          Artibeus jamaicensis aja-miR-21 MIMAT0031140
          Cricetulus griseus cgr-miR-21-3p MIMAT0022937
          Dasypus novemcinctus dno-miR-21-3p MIMAT0047525
          Eptesicus fuscus efu-miR-21 MIMAT0035146
          View More Rows
          Guinea pig cpo-miR-21-3p MIMAT0046873
          Ornithorhynchus anatinus oan-miR-21-3p MIMAT0007161
          Rabbit ocu-miR-21-3p MIMAT0048125

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-21a
          Stem-loop Accession # MI0000569
          Stem-loop Sequence
          UGUACCACCUUGUCGGAUAGCUUAUCAGACUGAUGUUGACUGUUGAAUCUCAUGGCAACAGCAGUCGAUGGGCUGUCUGACAUUUUGGUAUC
          Chromosome Location Chr. 11 - 86584067 - 86584158 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004628
          miRBase ID: mmu-miR-21a-3p
          Mature miRNA Sequence: CAACAGCAGUCGAUGGGCUGUC
          Chromosome Location: Chr. 11 - 86584067 - 86584158 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-21
          Stem-loop Accession # MI0000850
          Stem-loop Sequence
          UGUACCACCUUGUCGGGUAGCUUAUCAGACUGAUGUUGACUGUUGAAUCUCAUGGCAACAGCAGUCGAUGGGCUGUCUGACAUUUUGGUAUC
          Chromosome Location Chr. 10 - 73902210 - 73902301 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004711
          miRBase ID: rno-miR-21-3p
          Mature miRNA Sequence: CAACAGCAGUCGAUGGGCUGUC
          Chromosome Location: Chr. 10 - 73902210 - 73902301 [-] on Build Rnor_6.0

          Artibeus jamaicensis

          Stem-loop ID aja-mir-21
          Stem-loop Accession # MI0024362
          Stem-loop Sequence
          GUACCACCUUGUCGGGUAGCUUAUCAGACUGAUGUUGACUGUUGACUCUCAUGGCAACAGCAGUCGAUGGGCUGUCCGACAUUCUGGUAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031140
          miRBase ID: aja-miR-21
          Mature miRNA Sequence: CAACAGCAGUCGAUGGGCUGUC
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-21
          Stem-loop Accession # MI0005725
          Stem-loop Sequence
          UGUACCACCUUGUCGGGUAGCUUAUCAGACUGAUGUUGACUGUUGAAUCUCAUGGCAACAGCAGUCGAUGGGCUGUCUGACAUUUUGGUAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0022937
          miRBase ID: cgr-miR-21-3p
          Mature miRNA Sequence: CAACAGCAGUCGAUGGGCUGUC
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-21
          Stem-loop Accession # MI0038917
          Stem-loop Sequence
          UAGCUUAUCAGACUGAUGUUGACUGUUGAAUCUCAUGGCAACAGCAGUCGAUGGGCUGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047525
          miRBase ID: dno-miR-21-3p
          Mature miRNA Sequence: CAACAGCAGUCGAUGGGCUGUC
          Chromosome Location: NA

          Eptesicus fuscus

          Stem-loop ID efu-mir-21
          Stem-loop Accession # MI0028851
          Stem-loop Sequence
          CCUUGUCGGGUAGCUUAUCAGACUGAUGUUGACUGUUGAAUCUCAUGGCAACAGCAGUCGAUGGGCUGUCUGACAUUUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035146
          miRBase ID: efu-miR-21
          Mature miRNA Sequence: CAACAGCAGUCGAUGGGCUGUC
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-21
          Stem-loop Accession # MI0038573
          Stem-loop Sequence
          UAGCUUAUCAGACUGAUGUUGACUGUUGAAUCUCAUGGCAACAGCAGUCGAUGGGCUGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046873
          miRBase ID: cpo-miR-21-3p
          Mature miRNA Sequence: CAACAGCAGUCGAUGGGCUGUC
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-21
          Stem-loop Accession # MI0006883
          Stem-loop Sequence
          AUCUCCACGACUGGACCAUCCUAUCGGAUAGCUUAUCAGACUGAUGUUGACUGUUAGAUCUCCUGGCAACAGCAGUCGAUGGGCUGUCUGACAUUUUGGUAUCUGUCAUCCGAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007161
          miRBase ID: oan-miR-21-3p
          Mature miRNA Sequence: CAACAGCAGUCGAUGGGCUGUC
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-21
          Stem-loop Accession # MI0039233
          Stem-loop Sequence
          UAGCUUAUCAGACUGAUGUUGACUGUUGAAUCUCAUGGCAACAGCAGUCGAUGGGCUGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048125
          miRBase ID: ocu-miR-21-3p
          Mature miRNA Sequence: CAACAGCAGUCGAUGGGCUGUC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03464993_pri
          TaqMan™ Pri-miRNA Assay : Mm03306822_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM13039
          Ambion® Pre-miR™ miRNA Precursor : PM13039
          TaqMan™ MicroRNA Assay : 002493
          mirVana® miRNA inhibitor : MH13039
          mirVana® miRNA mimic : MC13039
          TaqMan™ Advanced miRNA Assay : rno480993_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-74676f44b6-kp6hx:80/100.66.72.150:80.
          git-commit: db542085ed6beb0c3f0dd774c2548ab49b9f1b6a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026.10.42-1.0