Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu481127_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-361-5p
          Stem-loop Accession Number: MI0000760
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 85903636 - 85903707 [-] on Build GRCh38
          Mature miRNA Sequence: UUAUCAGAAUCUCCAGGGGUAC
          Species: Human, Mouse, Rat, Bovine, Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Dog, Eptesicus fuscus, Goat, Gorilla gorilla, Guinea pig, Horse, Microcebus murinus, Nomascus leucogenys, Otolemur garnettii, Pig, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu481127_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000172, mir-361

          Mature miRNA Sequence:

          UUAUCAGAAUCUCCAGGGGUAC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-361-5p MIMAT0000703
          Mouse mmu-miR-361-5p MIMAT0000704
          Rat rno-miR-361-5p MIMAT0003117
          Bovine bta-miR-361 MIMAT0003830
          Callithrix jacchus cja-miR-361 MIMAT0039513
          Cricetulus griseus cgr-miR-361 MIMAT0023933
          View More Rows
          Dasypus novemcinctus dno-miR-361-5p MIMAT0047829
          Dog cfa-miR-361 MIMAT0006751
          Eptesicus fuscus efu-miR-361 MIMAT0035058
          Goat chi-miR-361-5p MIMAT0036171
          Gorilla gorilla ggo-miR-361 MIMAT0024136
          Guinea pig cpo-miR-361-5p MIMAT0047188
          Horse eca-miR-361-5p MIMAT0013208
          Microcebus murinus mmr-miR-361 MIMAT0049172
          Nomascus leucogenys nle-miR-361 MIMAT0049413
          Otolemur garnettii oga-miR-361 MIMAT0049682
          Pig ssc-miR-361-5p MIMAT0013933
          Pongo pygmaeus ppy-miR-361-5p MIMAT0015848
          Pteropus alecto pal-miR-361-5p MIMAT0040039
          Rabbit ocu-miR-361-5p MIMAT0048393
          Rhesus monkey mml-miR-361-5p MIMAT0006286

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-361
          Stem-loop Accession # MI0000760
          Stem-loop Sequence
          GGAGCUUAUCAGAAUCUCCAGGGGUACUUUAUAAUUUCAAAAAGUCCCCCAGGUGUGAUUCUGAUUUGCUUC
          Chromosome Location Chr. X - 85903636 - 85903707 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000703
          miRBase ID: hsa-miR-361-5p
          Mature miRNA Sequence: UUAUCAGAAUCUCCAGGGGUAC
          Chromosome Location: Chr. X - 85903636 - 85903707 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-361
          Stem-loop Accession # MI0000761
          Stem-loop Sequence
          GAAGCUUAUCAGAAUCUCCAGGGGUACUUAGUAUUUGAAAAGUCCCCCAGGUGUGAUUCUGAUUUGUUUC
          Chromosome Location Chr. X - 113074824 - 113074893 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000704
          miRBase ID: mmu-miR-361-5p
          Mature miRNA Sequence: UUAUCAGAAUCUCCAGGGGUAC
          Chromosome Location: Chr. X - 113074824 - 113074893 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-361
          Stem-loop Accession # MI0003481
          Stem-loop Sequence
          GAAGCUUAUCAGAAUCUCCAGGGGUACUUAUUAUUUGAAAAGUCCCCCAGGUGUGAUUCUGAUUCGUUUC
          Chromosome Location Chr. X - 84708911 - 84708980 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0003117
          miRBase ID: rno-miR-361-5p
          Mature miRNA Sequence: UUAUCAGAAUCUCCAGGGGUAC
          Chromosome Location: Chr. X - 84708911 - 84708980 [-] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-361
          Stem-loop Accession # MI0005045
          Stem-loop Sequence
          GGAGCUUAUCAGAAUCUCCAGGGGUACUUAUAAUUUGAAAAAGUCCCCCAGGUGUGAUUCUGAUUUGCUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003830
          miRBase ID: bta-miR-361
          Mature miRNA Sequence: UUAUCAGAAUCUCCAGGGGUAC
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-361
          Stem-loop Accession # MI0032040
          Stem-loop Sequence
          GCAGUGGCAUGCUUGACAGUGAUUCUUUCCUGGGAUUUGGGAGCUUAUCAGAAUCUCCAGGGGUACUUUAUAAUUUAAAAAUGUCCCCCAGGUGUGAUUCUGAUUUGCUUCCUUCUCCUCCCACUCUGACAAUCACUGUUUCAUUAGAAGCCUCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039513
          miRBase ID: cja-miR-361
          Mature miRNA Sequence: UUAUCAGAAUCUCCAGGGGUAC
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-361
          Stem-loop Accession # MI0020509
          Stem-loop Sequence
          CUUGGAAGCUUAUCAGAAUCUCCAGGGGUACUUAUUAUUUGAAAAAGUCCCCCAGGUGUGGUUCUGAUUCGUUUCCUUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023933
          miRBase ID: cgr-miR-361
          Mature miRNA Sequence: UUAUCAGAAUCUCCAGGGGUAC
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-361
          Stem-loop Accession # MI0039079
          Stem-loop Sequence
          UUAUCAGAAUCUCCAGGGGUACUUAAUAUGUGAAAAAAGUCCCCCAGGUGUGAUUCUGAUUUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047829
          miRBase ID: dno-miR-361-5p
          Mature miRNA Sequence: UUAUCAGAAUCUCCAGGGGUAC
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-361
          Stem-loop Accession # MI0008177
          Stem-loop Sequence
          UUAUCAGAAUCUCCAGGGGUACUUAAAAUUUGAAAAAGUCCCCCAGGUGUGAUUCUGAUUUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006751
          miRBase ID: cfa-miR-361
          Mature miRNA Sequence: UUAUCAGAAUCUCCAGGGGUAC
          Chromosome Location: NA

          Eptesicus fuscus

          Stem-loop ID efu-mir-361
          Stem-loop Accession # MI0028745
          Stem-loop Sequence
          AUUUGGGAGCUUAUCAGAAUCUCCAGGGGUACUUAUAAUUUGAAAAAGUCCCCCAGGUGUGAUUCUGAUUUGCUUCCUUCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035058
          miRBase ID: efu-miR-361
          Mature miRNA Sequence: UUAUCAGAAUCUCCAGGGGUAC
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-361
          Stem-loop Accession # MI0030760
          Stem-loop Sequence
          CCUGGGAUUUGGGAGCUUAUCAGAAUCUCCAGGGGUACUUAUAAUUUGAAAAAGUCCCCCAGGUGUGAUUCUGAUUUGCUUCCUUCUCCUCCUCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036171
          miRBase ID: chi-miR-361-5p
          Mature miRNA Sequence: UUAUCAGAAUCUCCAGGGGUAC
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-361
          Stem-loop Accession # MI0020683
          Stem-loop Sequence
          UUUUCCUGGGAUUUGGGAGCUUAUCAGAAUCUCCAGGGGUACUUUAUAAUUUCAAAAAGUCCCCCAGGUGUGAUUCUGAUUUGCUUCCUUUUCCUCCUCCUGCUCUGAGAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024136
          miRBase ID: ggo-miR-361
          Mature miRNA Sequence: UUAUCAGAAUCUCCAGGGGUAC
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-361
          Stem-loop Accession # MI0038742
          Stem-loop Sequence
          UUAUCAGAAUCUCCAGGGGUACUUAUAAUUUGUAAAAGUCCCCCAGGUGUGAUUCUGAUUUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047188
          miRBase ID: cpo-miR-361-5p
          Mature miRNA Sequence: UUAUCAGAAUCUCCAGGGGUAC
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-361
          Stem-loop Accession # MI0012956
          Stem-loop Sequence
          GGAGCUUAUCAGAAUCUCCAGGGGUACUGAUAAUUUUAAAAAGUCCCCCAGGCGUGAUUCUGAUUUGCUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013208
          miRBase ID: eca-miR-361-5p
          Mature miRNA Sequence: UUAUCAGAAUCUCCAGGGGUAC
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-361
          Stem-loop Accession # MI0039889
          Stem-loop Sequence
          UUAUCAGAAUCUCCAGGGGUACUUAUAAUUUGAAAAAGUCCCCCAGGUGUGAUUCUGAUUUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049172
          miRBase ID: mmr-miR-361
          Mature miRNA Sequence: UUAUCAGAAUCUCCAGGGGUAC
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-361
          Stem-loop Accession # MI0040154
          Stem-loop Sequence
          UUAUCAGAAUCUCCAGGGGUACUUUAUAAUUUCAAAAAGUCCCCCAGGUGUGAUUCUGAUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049413
          miRBase ID: nle-miR-361
          Mature miRNA Sequence: UUAUCAGAAUCUCCAGGGGUAC
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-361
          Stem-loop Accession # MI0040465
          Stem-loop Sequence
          UUAUCAGAAUCUCCAGGGGUACUUAUAAUUUGAAAAAGUCCCCCAGGUGUGAUUCUGAUUUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049682
          miRBase ID: oga-miR-361
          Mature miRNA Sequence: UUAUCAGAAUCUCCAGGGGUAC
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-361
          Stem-loop Accession # MI0013145
          Stem-loop Sequence
          GGAGCUUAUCAGAAUCUCCAGGGGUACUUAUAAUUUGAAAAAGUCCCCCAGGUGUGAUUCUGAUUUGCUUCCUCCUCCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013933
          miRBase ID: ssc-miR-361-5p
          Mature miRNA Sequence: UUAUCAGAAUCUCCAGGGGUAC
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-361
          Stem-loop Accession # MI0014908
          Stem-loop Sequence
          GGAGCUUAUCAGAAUCUCCAGGGGUACUUUAUAAUUUCAAAAAGUCCCCCAGGUGUGAUUCUGAUUUGCUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015848
          miRBase ID: ppy-miR-361-5p
          Mature miRNA Sequence: UUAUCAGAAUCUCCAGGGGUAC
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-361
          Stem-loop Accession # MI0032462
          Stem-loop Sequence
          UUAUCAGAAUCUCCAGGGGUACUUAUAAUUUGAAAAAGUCCCCCAGGUGCGAUUCUGAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040039
          miRBase ID: pal-miR-361-5p
          Mature miRNA Sequence: UUAUCAGAAUCUCCAGGGGUAC
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-361
          Stem-loop Accession # MI0039377
          Stem-loop Sequence
          UUAUCAGAAUCUCCAGGGGUACUUACAAUUUGAAAAAGUCCCCCAGGUGUGAUUCUGAUUUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048393
          miRBase ID: ocu-miR-361-5p
          Mature miRNA Sequence: UUAUCAGAAUCUCCAGGGGUAC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-361
          Stem-loop Accession # MI0007707
          Stem-loop Sequence
          GGAGCUUAUCAGAAUCUCCAGGGGUACUUUAUAAUUUCAAAAAGUCCCCCAGGUGUGAUUCUGAUUUGCUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006286
          miRBase ID: mml-miR-361-5p
          Mature miRNA Sequence: UUAUCAGAAUCUCCAGGGGUAC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303437_pri
          TaqMan™ Pri-miRNA Assay : Mm03307318_pri
          TaqMan™ Pri-miRNA Assay : Rn03465512_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10085
          Ambion® Pre-miR™ miRNA Precursor : PM10085
          TaqMan™ MicroRNA Assay : 000554
          mirVana® miRNA inhibitor : MH10085
          mirVana® miRNA mimic : MC10085
          TaqMan™ Advanced miRNA Assay : 478056_mir
          TaqMan™ Advanced miRNA Assay : rno481127_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline