Search
Search

| Assay Name: | mmu-miR-93-3p |
|---|---|
| Stem-loop Accession Number: | MI0000095 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | ACUGCUGAGCUAGCACUUCCCG |
| Species: | Human, Mouse, Pteropus alecto, Rhesus monkey |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000001, mir-17 |
Mature miRNA Sequence: | ACUGCUGAGCUAGCACUUCCCG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-93-3p | MIMAT0004509 |
| Mouse | mmu-miR-93-3p | MIMAT0004636 |
| Pteropus alecto | pal-miR-93-3p | MIMAT0039938 |
| Rhesus monkey | mml-miR-93-3p | MIMAT0026602 |