Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu481343_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-210-3p
          Stem-loop Accession Number: MI0000286
          miRBase Version: v22.1
          Chromosome Location: Chr.11: 568089 - 568198 [-] on Build GRCh38
          Mature miRNA Sequence: CUGUGCGUGUGACAGCGGCUGA
          Species: Human, Mouse, Rat, Branchiostoma belcheri, Branchiostoma floridae, Cricetulus griseus, Dasypus novemcinctus, Gorilla gorilla, Guinea pig, Nomascus leucogenys, Otolemur garnettii, Pan paniscus, Pan troglodytes, Papio hamadryas, Pig, Rhesus monkey, Tupaia chinensis
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu481343_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000086, mir-210

          Mature miRNA Sequence:

          CUGUGCGUGUGACAGCGGCUGA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-210-3p MIMAT0000267
          Mouse mmu-miR-210-3p MIMAT0000658
          Rat rno-miR-210-3p MIMAT0000881
          Branchiostoma belcheri bbe-miR-210-5p MIMAT0031645
          Branchiostoma floridae bfl-miR-210-3p MIMAT0009494
          Cricetulus griseus cgr-miR-210-3p MIMAT0023849
          View More Rows
          Dasypus novemcinctus dno-miR-210-3p MIMAT0047755
          Gorilla gorilla ggo-miR-210 MIMAT0024146
          Guinea pig cpo-miR-210-3p MIMAT0047110
          Nomascus leucogenys nle-miR-210 MIMAT0049339
          Otolemur garnettii oga-miR-210 MIMAT0049660
          Pan paniscus ppa-miR-210 MIMAT0049101
          Pan troglodytes ptr-miR-210 MIMAT0008070
          Papio hamadryas pha-miR-210 MIMAT0049575
          Pig ssc-miR-210 MIMAT0007761
          Rhesus monkey mml-miR-210-3p MIMAT0006240
          Tupaia chinensis tch-miR-210 MIMAT0036464

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-210
          Stem-loop Accession # MI0000286
          Stem-loop Sequence
          ACCCGGCAGUGCCUCCAGGCGCAGGGCAGCCCCUGCCCACCGCACACUGCGCUGCCCCAGACCCACUGUGCGUGUGACAGCGGCUGAUCUGUGCCUGGGCAGCGCGACCC
          Chromosome Location Chr. 11 - 568089 - 568198 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000267
          miRBase ID: hsa-miR-210-3p
          Mature miRNA Sequence: CUGUGCGUGUGACAGCGGCUGA
          Chromosome Location: Chr. 11 - 568089 - 568198 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-210
          Stem-loop Accession # MI0000695
          Stem-loop Sequence
          CCGGGGCAGUCCCUCCAGGCUCAGGACAGCCACUGCCCACCGCACACUGCGUUGCUCCGGACCCACUGUGCGUGUGACAGCGGCUGAUCUGUCCCUGGGCAGCGCGAACC
          Chromosome Location Chr. 7 - 141221384 - 141221493 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000658
          miRBase ID: mmu-miR-210-3p
          Mature miRNA Sequence: CUGUGCGUGUGACAGCGGCUGA
          Chromosome Location: Chr. 7 - 141221384 - 141221493 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-210
          Stem-loop Accession # MI0000950
          Stem-loop Sequence
          CCGGGGCAGUCCCUCCAGGCUCAGGACAGCCACUGCCCACAGCACACUGCGUUGCUCCGGACCCACUGUGCGUGUGACAGCGGCUGAUCUGUCCCUGGGCAGCGCGAACC
          Chromosome Location Chr. 1 - 214208355 - 214208464 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000881
          miRBase ID: rno-miR-210-3p
          Mature miRNA Sequence: CUGUGCGUGUGACAGCGGCUGA
          Chromosome Location: Chr. 1 - 214208355 - 214208464 [-] on Build Rnor_6.0

          Branchiostoma belcheri

          Stem-loop ID bbe-mir-210
          Stem-loop Accession # MI0026220
          Stem-loop Sequence
          GUAGUUGGUCGGUGGCAUUCGCCUCUGUUUCGCGCGCAAGAUGUGAUGUCAGUCCUUCUCCUGUGCGUGUGACAGCGGCUGAUGCACCGGCCAAAAAGUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031645
          miRBase ID: bbe-miR-210-5p
          Mature miRNA Sequence: CUGUGCGUGUGACAGCGGCUGA
          Chromosome Location: NA

          Branchiostoma floridae

          Stem-loop ID bfl-mir-210
          Stem-loop Accession # MI0010038
          Stem-loop Sequence
          ACAGUUGGUCGGUGGCAUUCGCCUCUGUUUCGCGCGCAAGAUGUGAUGUUAGUCCUGCUCCUGUGCGUGUGACAGCGGCUGAUGCACCGGCCAAAACGUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009494
          miRBase ID: bfl-miR-210-3p
          Mature miRNA Sequence: CUGUGCGUGUGACAGCGGCUGA
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-210
          Stem-loop Accession # MI0020458
          Stem-loop Sequence
          GGCUCAGGACAGCCACUGCCCACCGCACACUGCGUUGCUCCGGACCCACUGUGCGUGUGACAGCGGCUGAUCUGUCCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023849
          miRBase ID: cgr-miR-210-3p
          Mature miRNA Sequence: CUGUGCGUGUGACAGCGGCUGA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-210
          Stem-loop Accession # MI0039041
          Stem-loop Sequence
          AGCCCCUGCCCACCGCACACUGCGCUGCUCCGGACCCACUGUGCGUGUGACAGCGGCUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047755
          miRBase ID: dno-miR-210-3p
          Mature miRNA Sequence: CUGUGCGUGUGACAGCGGCUGA
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-210
          Stem-loop Accession # MI0020693
          Stem-loop Sequence
          GCCCACCCGGCAGUCCCUCCAGGCGCAGGGCAGCCCCUGCCCACCGCACACUGCGCUGCCCCAGACCCACUGUGCGUGUGACAGCGGCUGAUCUGUGCCUGGGCAGCGCGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024146
          miRBase ID: ggo-miR-210
          Mature miRNA Sequence: CUGUGCGUGUGACAGCGGCUGA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-210
          Stem-loop Accession # MI0038702
          Stem-loop Sequence
          GCGCAGGGCAGCCACUGCCCACCGCACACUGCGCUGCUCCCGACCCACUGUGCGUGUGACAGCGGCUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047110
          miRBase ID: cpo-miR-210-3p
          Mature miRNA Sequence: CUGUGCGUGUGACAGCGGCUGA
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-210
          Stem-loop Accession # MI0040075
          Stem-loop Sequence
          AGCCCCUGCCCACCGCACACUGCGCUGCCCCAGACCCACUGUGCGUGUGACAGCGGCUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049339
          miRBase ID: nle-miR-210
          Mature miRNA Sequence: CUGUGCGUGUGACAGCGGCUGA
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-210
          Stem-loop Accession # MI0040443
          Stem-loop Sequence
          AGCCACUGCCCACCGCACACUGCGCUGCUCCAGACCCACUGUGCGUGUGACAGCGGCUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049660
          miRBase ID: oga-miR-210
          Mature miRNA Sequence: CUGUGCGUGUGACAGCGGCUGA
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-210
          Stem-loop Accession # MI0039813
          Stem-loop Sequence
          AGCCCCUGCCCACCGCACACUGCGCUGCCCCAGACCCACUGUGCGUGUGACAGCGGCUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049101
          miRBase ID: ppa-miR-210
          Mature miRNA Sequence: CUGUGCGUGUGACAGCGGCUGA
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-210
          Stem-loop Accession # MI0008580
          Stem-loop Sequence
          ACCCGGCAGUCCCUCCAGGCGCAGGGCAGCCCCUGCCCACCGCACACUGCGCUGCCCCAGACCCACUGUGCGUGUGACAGCGGCUGAUCUGUGCCUGGGCAGCGCGACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008070
          miRBase ID: ptr-miR-210
          Mature miRNA Sequence: CUGUGCGUGUGACAGCGGCUGA
          Chromosome Location: NA

          Papio hamadryas

          Stem-loop ID pha-mir-210-1
          Stem-loop Accession # MI0040350
          Stem-loop Sequence
          AGCCCCUGCCCACCGCACACUGCGCUGCCCCAGACCCACUGUGCGUGUGACAGCGGCUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049575
          miRBase ID: pha-miR-210
          Mature miRNA Sequence: CUGUGCGUGUGACAGCGGCUGA
          Chromosome Location: NA

          Stem-loop ID pha-mir-210-2
          Stem-loop Accession # MI0040373
          Stem-loop Sequence
          AGCCCCUGCCCACCGCACACUGCGCUGCCCCAGACCCACUGUGCGUGUGACAGCGGCUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049575
          miRBase ID: pha-miR-210
          Mature miRNA Sequence: CUGUGCGUGUGACAGCGGCUGA
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-210
          Stem-loop Accession # MI0008220
          Stem-loop Sequence
          GCGCAGGGCAGCCACUGCCCACCGCACACUGCGCUGCUCCGGACCCACUGUGCGUGUGACAGCGGCUGAUCUGUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007761
          miRBase ID: ssc-miR-210
          Mature miRNA Sequence: CUGUGCGUGUGACAGCGGCUGA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-210
          Stem-loop Accession # MI0007670
          Stem-loop Sequence
          ACCCGGCAGUCCCUCCAGGCGCAGGGCAGCCCCUGCCCACCGCACACUGCGCUGCCCCAGACCCACUGUGCGUGUGACAGCGGCUGAUCUGUGCCUGGGCAGCGCGACCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006240
          miRBase ID: mml-miR-210-3p
          Mature miRNA Sequence: CUGUGCGUGUGACAGCGGCUGA
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-210
          Stem-loop Accession # MI0031123
          Stem-loop Sequence
          AGCCACUGCCCACCGCACACUGCGCUGCUCCGGACCCACUGUGCGUGUGACAGCGGCUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036464
          miRBase ID: tch-miR-210
          Mature miRNA Sequence: CUGUGCGUGUGACAGCGGCUGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302948_pri
          TaqMan™ Pri-miRNA Assay : Mm03307104_pri
          TaqMan™ Pri-miRNA Assay : Rn03466077_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10516
          Ambion® Pre-miR™ miRNA Precursor : PM10516
          TaqMan™ MicroRNA Assay : 000512
          mirVana® miRNA inhibitor : MH10516
          mirVana® miRNA mimic : MC10516
          TaqMan™ Advanced miRNA Assay : 477970_mir
          TaqMan™ Advanced miRNA Assay : rno481343_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch