Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu481511_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-let-7i-3p
          Stem-loop Accession Number: MI0000434
          miRBase Version: v22.1
          Chromosome Location: Chr.12: 62603686 - 62603769 [+] on Build GRCh38
          Mature miRNA Sequence: CUGCGCAAGCUACUGCCUUGCU
          Species: Human, Mouse, Rat, Alligator mississippiensis, Artibeus jamaicensis, Chrysemys picta, Columba livia, Dasypus novemcinctus, Goat, Guinea pig, Pig, Python bivittatus, Rabbit, Taeniopygia guttata
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu481511_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000002, let-7

          Mature miRNA Sequence:

          CUGCGCAAGCUACUGCCUUGCU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-let-7i-3p MIMAT0004585
          Mouse mmu-let-7i-3p MIMAT0004520
          Rat rno-let-7i-3p MIMAT0004707
          Alligator mississippiensis ami-let-7i-3p MIMAT0038017
          Artibeus jamaicensis aja-let-7i MIMAT0031151
          Chrysemys picta cpi-let-7i-3p MIMAT0037611
          View More Rows
          Columba livia cli-let-7i-3p MIMAT0038390
          Dasypus novemcinctus dno-let-7i-3p MIMAT0047487
          Goat chi-let-7i-3p MIMAT0035895
          Guinea pig cpo-let-7i-3p MIMAT0046834
          Pig ssc-let-7i-3p MIMAT0037319
          Python bivittatus pbv-let-7i-3p MIMAT0038811
          Rabbit ocu-let-7i-3p MIMAT0048090
          Taeniopygia guttata tgu-let-7i-3p MIMAT0026974

          Stem-loop Details



          Human

          Stem-loop ID hsa-let-7i
          Stem-loop Accession # MI0000434
          Stem-loop Sequence
          CUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUA
          Chromosome Location Chr. 12 - 62603686 - 62603769 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004585
          miRBase ID: hsa-let-7i-3p
          Mature miRNA Sequence: CUGCGCAAGCUACUGCCUUGCU
          Chromosome Location: Chr. 12 - 62603686 - 62603769 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-let-7i
          Stem-loop Accession # MI0000138
          Stem-loop Sequence
          CUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUAG
          Chromosome Location Chr. 10 - 122985640 - 122985724 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004520
          miRBase ID: mmu-let-7i-3p
          Mature miRNA Sequence: CUGCGCAAGCUACUGCCUUGCU
          Chromosome Location: Chr. 10 - 122985640 - 122985724 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-let-7i
          Stem-loop Accession # MI0000835
          Stem-loop Sequence
          CUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUAG
          Chromosome Location Chr. 7 - 66802731 - 66802815 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004707
          miRBase ID: rno-let-7i-3p
          Mature miRNA Sequence: CUGCGCAAGCUACUGCCUUGCU
          Chromosome Location: Chr. 7 - 66802731 - 66802815 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-let-7i
          Stem-loop Accession # MI0029651
          Stem-loop Sequence
          CGGUCCUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACACUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUAGUGCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038017
          miRBase ID: ami-let-7i-3p
          Mature miRNA Sequence: CUGCGCAAGCUACUGCCUUGCU
          Chromosome Location: NA

          Artibeus jamaicensis

          Stem-loop ID aja-let-7i
          Stem-loop Accession # MI0024373
          Stem-loop Sequence
          CUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031151
          miRBase ID: aja-let-7i
          Mature miRNA Sequence: CUGCGCAAGCUACUGCCUUGCU
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-let-7i
          Stem-loop Accession # MI0029383
          Stem-loop Sequence
          CCCUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUAGUGCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037611
          miRBase ID: cpi-let-7i-3p
          Mature miRNA Sequence: CUGCGCAAGCUACUGCCUUGCU
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-let-7i
          Stem-loop Accession # MI0029892
          Stem-loop Sequence
          CUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038390
          miRBase ID: cli-let-7i-3p
          Mature miRNA Sequence: CUGCGCAAGCUACUGCCUUGCU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-let-7i
          Stem-loop Accession # MI0038894
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047487
          miRBase ID: dno-let-7i-3p
          Mature miRNA Sequence: CUGCGCAAGCUACUGCCUUGCU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-let-7i
          Stem-loop Accession # MI0030586
          Stem-loop Sequence
          CCCGACACCAUGGCCCUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUAGUGCUGGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035895
          miRBase ID: chi-let-7i-3p
          Mature miRNA Sequence: CUGCGCAAGCUACUGCCUUGCU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-let-7i
          Stem-loop Accession # MI0038550
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046834
          miRBase ID: cpo-let-7i-3p
          Mature miRNA Sequence: CUGCGCAAGCUACUGCCUUGCU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-let-7i
          Stem-loop Accession # MI0002447
          Stem-loop Sequence
          CUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037319
          miRBase ID: ssc-let-7i-3p
          Mature miRNA Sequence: CUGCGCAAGCUACUGCCUUGCU
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-let-7i
          Stem-loop Accession # MI0030142
          Stem-loop Sequence
          ACCCUGGCUGAGGUAGUAGUUUGUGCUGUUCGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUAGUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038811
          miRBase ID: pbv-let-7i-3p
          Mature miRNA Sequence: CUGCGCAAGCUACUGCCUUGCU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-let-7i
          Stem-loop Accession # MI0039214
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048090
          miRBase ID: ocu-let-7i-3p
          Mature miRNA Sequence: CUGCGCAAGCUACUGCCUUGCU
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-let-7i
          Stem-loop Accession # MI0013694
          Stem-loop Sequence
          GGGCCCUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUAGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026974
          miRBase ID: tgu-let-7i-3p
          Mature miRNA Sequence: CUGCGCAAGCUACUGCCUUGCU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306160_pri
          TaqMan™ Pri-miRNA Assay : Rn03465226_pri
          TaqMan™ Pri-miRNA Assay : Hs03303039_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12783
          Ambion® Pre-miR™ miRNA Precursor : PM12783
          TaqMan™ MicroRNA Assay : 002172
          mirVana® miRNA inhibitor : MH12783
          mirVana® miRNA mimic : MC12783
          TaqMan™ Advanced miRNA Assay : rno480880_mir
          TaqMan™ Advanced miRNA Assay : 477862_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline