Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu482714_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-23b-5p
          Stem-loop Accession Number: MI0000141
          miRBase Version: v22.1
          Chromosome Location: Chr.13: 63300484 - 63300557 [+] on Build GRCm38
          Mature miRNA Sequence: GGGUUCCUGGCAUGCUGAUUU
          Species: Mouse, Rat, Alligator mississippiensis, Anolis carolinensis, Bovine, Dasypus novemcinctus, Guinea pig, Monodelphis domestica, Pteropus alecto, Python bivittatus, Rabbit, Rhesus monkey
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu482714_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000027, mir-23

          Mature miRNA Sequence:

          GGGUUCCUGGCAUGCUGAUUU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-23b-5p MIMAT0016980
          Rat rno-miR-23b-5p MIMAT0017099
          Alligator mississippiensis ami-miR-23b-5p MIMAT0038059
          Anolis carolinensis aca-miR-23b-5p MIMAT0021897
          Bovine bta-miR-23b-5p MIMAT0012538
          Dasypus novemcinctus dno-miR-23b-5p MIMAT0047530
          View More Rows
          Guinea pig cpo-miR-23b-5p MIMAT0046878
          Monodelphis domestica mdo-miR-23b-5p MIMAT0026713
          Pteropus alecto pal-miR-23b-5p MIMAT0039870
          Python bivittatus pbv-miR-23b-5p MIMAT0038854
          Rabbit ocu-miR-23b-5p MIMAT0048128
          Rhesus monkey mml-miR-23b-5p MIMAT0026800

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-23b
          Stem-loop Accession # MI0000141
          Stem-loop Sequence
          GGCUGCUUGGGUUCCUGGCAUGCUGAUUUGUGACUUGAGAUUAAAAUCACAUUGCCAGGGAUUACCACGCAACC
          Chromosome Location Chr. 13 - 63300484 - 63300557 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0016980
          miRBase ID: mmu-miR-23b-5p
          Mature miRNA Sequence: GGGUUCCUGGCAUGCUGAUUU
          Chromosome Location: Chr. 13 - 63300484 - 63300557 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-23b
          Stem-loop Accession # MI0000853
          Stem-loop Sequence
          CUCACCUGCUCUGGCUGCUUGGGUUCCUGGCAUGCUGAUUUGUGACUUGAGAUUAAAAUCACAUUGCCAGGGAUUACCACGCAACCAUGACCUUGGC
          Chromosome Location Chr. 17 - 823222 - 823318 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017099
          miRBase ID: rno-miR-23b-5p
          Mature miRNA Sequence: GGGUUCCUGGCAUGCUGAUUU
          Chromosome Location: Chr. 17 - 823222 - 823318 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-23b
          Stem-loop Accession # MI0029679
          Stem-loop Sequence
          CCCAGUGUUGUGGCUGUUUGGGUUCCUGGCAUGCUGAUUUGUGACUUAAAAUUAAAAUCACAUUGCCAGGGAUUACCACAGAGCCAUGACCAUGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038059
          miRBase ID: ami-miR-23b-5p
          Mature miRNA Sequence: GGGUUCCUGGCAUGCUGAUUU
          Chromosome Location: NA

          Anolis carolinensis

          Stem-loop ID aca-mir-23b
          Stem-loop Accession # MI0018832
          Stem-loop Sequence
          GUUUUGGUUGUUUGGGUUCCUGGCAUGCUGAUUUGUGACUUAAGAUGAAAAUCACAUUGCCAGGGAUUACCACAUAACCAUGACAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021897
          miRBase ID: aca-miR-23b-5p
          Mature miRNA Sequence: GGGUUCCUGGCAUGCUGAUUU
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-23b
          Stem-loop Accession # MI0005066
          Stem-loop Sequence
          GGGUUCCUGGCAUGCUGAUUUGUGACUUAAGAUUAAAAUCACAUUGCCAGGGAUUACCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012538
          miRBase ID: bta-miR-23b-5p
          Mature miRNA Sequence: GGGUUCCUGGCAUGCUGAUUU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-23b
          Stem-loop Accession # MI0038920
          Stem-loop Sequence
          GGGUUCCUGGCAUGCUGAUUUGUGACUUAAGAUUAAAAUCACAUUGCCAGGGAUUACCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047530
          miRBase ID: dno-miR-23b-5p
          Mature miRNA Sequence: GGGUUCCUGGCAUGCUGAUUU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-23b
          Stem-loop Accession # MI0038576
          Stem-loop Sequence
          GGGUUCCUGGCAUGCUGAUUUGUGACUUAAGAUUAAAAUCACAUUGCCAGGGAUUACCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046878
          miRBase ID: cpo-miR-23b-5p
          Mature miRNA Sequence: GGGUUCCUGGCAUGCUGAUUU
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-23b
          Stem-loop Accession # MI0005366
          Stem-loop Sequence
          GGCUGCUUGGGUUCCUGGCAUGCUGAUUUGUGACUUAAGAUGAAAAUCACAUUGCCAGGGAUUACCACGCAGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026713
          miRBase ID: mdo-miR-23b-5p
          Mature miRNA Sequence: GGGUUCCUGGCAUGCUGAUUU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-23b
          Stem-loop Accession # MI0032373
          Stem-loop Sequence
          GGGUUCCUGGCAUGCUGAUUUGUGACUUAAGAUUAAAAUCACAUUGCCAGGGAUUACCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039870
          miRBase ID: pal-miR-23b-5p
          Mature miRNA Sequence: GGGUUCCUGGCAUGCUGAUUU
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-23b
          Stem-loop Accession # MI0030171
          Stem-loop Sequence
          CUGUUUUGGUUGUUUGGGUUCCUGGCAUGCUGAUUUGUGAUUUAAGAUUAAAAUCACAUUGCCAGGGAUUACCACAUAACCAUGACAUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038854
          miRBase ID: pbv-miR-23b-5p
          Mature miRNA Sequence: GGGUUCCUGGCAUGCUGAUUU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-23b
          Stem-loop Accession # MI0039235
          Stem-loop Sequence
          GGGUUCCUGGCAUGCUGAUUUGUGACUUAAGAUUAAAAUCACAUUGCCAGGGAUUACCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048128
          miRBase ID: ocu-miR-23b-5p
          Mature miRNA Sequence: GGGUUCCUGGCAUGCUGAUUU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-23b
          Stem-loop Accession # MI0007592
          Stem-loop Sequence
          CUCAGGUGCUCUGGCUGCUUGGGUUCCUGGCAUGCUGAUUUGUGACUUAAGAUUAAAAUCACAUUGCCAGGGAUUACCACGCAACCACGACCUUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026800
          miRBase ID: mml-miR-23b-5p
          Mature miRNA Sequence: GGGUUCCUGGCAUGCUGAUUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306184_pri
          TaqMan™ Pri-miRNA Assay : Rn03465744_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM15655
          Ambion® Pre-miR™ miRNA Precursor : PM15655
          TaqMan™ MicroRNA Assay : 243680_mat
          mirVana® miRNA inhibitor : MH15655
          mirVana® miRNA mimic : MC15655
          TaqMan™ Advanced miRNA Assay : rno481351_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline