Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu483162_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-let-7a-1-3p
          Stem-loop Accession Number: MI0000556
          miRBase Version: v22.1
          Chromosome Location: Chr.13: 48538179 - 48538272 [-] on Build GRCm38
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUCC
          Species: Mouse, Rat, Chicken, Columba livia, Dasypus novemcinctus, Gadus morhua, Goat, Guinea pig, Ornithorhynchus anatinus, Pteropus alecto, Python bivittatus, Rabbit, Rhesus monkey, Salmo salar, Taeniopygia guttata, Tupaia chinensis, Xenopus laevis
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu483162_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000002, let-7

          Mature miRNA Sequence:

          CUAUACAAUCUACUGUCUUUCC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-let-7a-1-3p MIMAT0004620
          Mouse mmu-let-7c-2-3p MIMAT0005439
          Rat rno-let-7a-1-3p MIMAT0017085
          Rat rno-let-7c-2-3p MIMAT0017088
          Chicken gga-let-7a-3p MIMAT0026491
          Chicken gga-let-7k-3p MIMAT0026543
          View More Rows
          Columba livia cli-let-7e-3p MIMAT0038384
          Dasypus novemcinctus dno-let-7a-3p MIMAT0047474
          Gadus morhua gmo-let-7e-3p MIMAT0044180
          Goat chi-let-7a-3p MIMAT0035881
          Guinea pig cpo-let-7a-3p MIMAT0046818
          Ornithorhynchus anatinus oan-let-7e-3p MIMAT0006985
          Pteropus alecto pal-let-7a-3p MIMAT0039773
          Python bivittatus pbv-let-7a-2-3p MIMAT0038795
          Python bivittatus pbv-let-7e-3p MIMAT0038804
          Python bivittatus pbv-let-7f-3p MIMAT0038806
          Rabbit ocu-let-7a-3p MIMAT0048076
          Rhesus monkey mml-let-7a-3p MIMAT0026789
          Salmo salar ssa-let-7e-3p MIMAT0032706
          Taeniopygia guttata tgu-let-7a-1-3p MIMAT0031085
          Taeniopygia guttata tgu-let-7e-3p MIMAT0026982
          Tupaia chinensis tch-let-7a-3p MIMAT0036484
          Xenopus laevis xla-let-7e-3p MIMAT0046402

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-let-7a-1
          Stem-loop Accession # MI0000556
          Stem-loop Sequence
          UUCACUGUGGGAUGAGGUAGUAGGUUGUAUAGUUUUAGGGUCACACCCACCACUGGGAGAUAACUAUACAAUCUACUGUCUUUCCUAAGGUGAU
          Chromosome Location Chr. 13 - 48538179 - 48538272 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004620
          miRBase ID: mmu-let-7a-1-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUCC
          Chromosome Location: Chr. 13 - 48538179 - 48538272 [-] on Build GRCm38

          Stem-loop ID mmu-let-7c-2
          Stem-loop Accession # MI0000560
          Stem-loop Sequence
          ACGGCCUUUGGGGUGAGGUAGUAGGUUGUAUGGUUUUGGGCUCUGCCCCGCUCUGCGGUAACUAUACAAUCUACUGUCUUUCCUGAAGUGGCCGC
          Chromosome Location Chr. 15 - 85706603 - 85706697 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0005439
          miRBase ID: mmu-let-7c-2-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUCC
          Chromosome Location: Chr. 15 - 85706603 - 85706697 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-let-7a-1
          Stem-loop Accession # MI0000827
          Stem-loop Sequence
          UUCACUGUGGGAUGAGGUAGUAGGUUGUAUAGUUUUAGGGUCACACCCACCACUGGGAGAUAACUAUACAAUCUACUGUCUUUCCUAAGGUGAU
          Chromosome Location Chr. 17 - 16417853 - 16417946 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017085
          miRBase ID: rno-let-7a-1-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUCC
          Chromosome Location: Chr. 17 - 16417853 - 16417946 [+] on Build Rnor_6.0

          Stem-loop ID rno-let-7c-2
          Stem-loop Accession # MI0000831
          Stem-loop Sequence
          ACGGCCUUUGGGGUGAGGUAGUAGGUUGUAUGGUUUUGGGCUCUGCCCCGCUCUGCGGUAACUAUACAAUCUACUGUCUUUCCUGAAGUGGCCGC
          Chromosome Location Chr. 7 - 126590212 - 126590306 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017088
          miRBase ID: rno-let-7c-2-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUCC
          Chromosome Location: Chr. 7 - 126590212 - 126590306 [+] on Build Rnor_6.0

          Chicken

          Stem-loop ID gga-let-7a-3
          Stem-loop Accession # MI0001171
          Stem-loop Sequence
          GGGUGAGGUAGUAGGUUGUAUAGUUUUAGGGUUAUGCCCUGCCUGUCAGAUAACUAUACAAUCUACUGUCUUUCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026491
          miRBase ID: gga-let-7a-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUCC
          Chromosome Location: NA

          Stem-loop ID gga-let-7k
          Stem-loop Accession # MI0001263
          Stem-loop Sequence
          GGCUGAGGUAGUAGAUUGAAUAGUUGUGGAGUCCUAUCCUCCCUUUGAGCUAACUAUACAAUCUACUGUCUUUCCUAAGGAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026543
          miRBase ID: gga-let-7k-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUCC
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-let-7e
          Stem-loop Accession # MI0029889
          Stem-loop Sequence
          CUGUCCUUGAGGCUGAGGUAGUAGAUUGAAUAGUUGUGGAGUCCGAUCCUCCCUUUGAGCAAACUAUACAAUCUACUGUCUUUCCUAAGGAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038384
          miRBase ID: cli-let-7e-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUCC
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-let-7a
          Stem-loop Accession # MI0038886
          Stem-loop Sequence
          UGAGGUAGUAGGUUGUAUAGUUUUAGGGUCACACCCACCACUGGGAGAUAACUAUACAAUCUACUGUCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047474
          miRBase ID: dno-let-7a-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUCC
          Chromosome Location: NA

          Gadus morhua

          Stem-loop ID gmo-let-7e-1
          Stem-loop Accession # MI0036099
          Stem-loop Sequence
          UGAGGUAGUAGAUUGAAUAGUUGUGGGGUUGUGCACCCCAAUCUGAGAUAACUAUACAAUCUACUGUCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044180
          miRBase ID: gmo-let-7e-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUCC
          Chromosome Location: NA

          Stem-loop ID gmo-let-7e-2
          Stem-loop Accession # MI0037979
          Stem-loop Sequence
          UGAGGUAGUAGAUUGAAUAGUUGUGGGGCCGAGUCCUCCCUCUGCGUUAACUAUACAAUCUACUGUCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044180
          miRBase ID: gmo-let-7e-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUCC
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-let-7a
          Stem-loop Accession # MI0030579
          Stem-loop Sequence
          AGACUGACGGCCCUUUGGGGUGAGGUAGUAGGUUGUAUAGUUUGGGGCUCUGCCCUGCUAUGGGAUAACUAUACAAUCUACUGUCUUUCCUGAAGUGGCUGUAAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035881
          miRBase ID: chi-let-7a-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUCC
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-let-7a-1
          Stem-loop Accession # MI0038540
          Stem-loop Sequence
          UGAGGUAGUAGGUUGUAUAGUUUUAGGGUCACACCCACCACUGGGAGAUAACUAUACAAUCUACUGUCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046818
          miRBase ID: cpo-let-7a-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUCC
          Chromosome Location: NA

          Stem-loop ID cpo-let-7a-3
          Stem-loop Accession # MI0038542
          Stem-loop Sequence
          UGAGGUAGUAGGUUGUAUAGUUUGGGGCUCCGCCCUGCUGUGGCUAACUAUACAAUCUACUGUCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046818
          miRBase ID: cpo-let-7a-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUCC
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-let-7e
          Stem-loop Accession # MI0006773
          Stem-loop Sequence
          GGCUCCCACCACAGUCCUGUCCUUGAGGCUGAGGUAGUAGAUUGAAUAGUUGUGGGGUCACGGCCUCCCUGUGUGCUAACUAUACAAUCUACUGUCUUUCCUGAGGCGACAGUCAGUUCGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006985
          miRBase ID: oan-let-7e-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUCC
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-let-7a-1
          Stem-loop Accession # MI0032320
          Stem-loop Sequence
          UGAGGUAGUAGGUUGUAUAGUUUGGGGCUCUGCCCUGCUAUGGGAUAACUAUACAAUCUACUGUCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039773
          miRBase ID: pal-let-7a-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUCC
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-let-7a-2
          Stem-loop Accession # MI0030131
          Stem-loop Sequence
          UGUCCUUUGGGGUGAGGUAGUAGGUUGUAUAGUUUUAGGGUUAGACCCUGCCUGUCAGAUAACUAUACAAUCUACUGUCUUUCCUGAAGUGGCUGUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038795
          miRBase ID: pbv-let-7a-2-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUCC
          Chromosome Location: NA

          Stem-loop ID pbv-let-7e
          Stem-loop Accession # MI0030138
          Stem-loop Sequence
          AUGCUGUCCUUGAGGCUGAGGUAGUAGAUUGAAUAGUUGUGGAGCCAUCUUCUCCCUUUGAGCUAACUAUACAAUCUACUGUCUUUCCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038804
          miRBase ID: pbv-let-7e-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUCC
          Chromosome Location: NA

          Stem-loop ID pbv-let-7f-1
          Stem-loop Accession # MI0030139
          Stem-loop Sequence
          CCUGGGAUGAGGUAGUAGAUUGUAUAGUUUUAGGGUCAUACCCGGUCUUGGAGAUAACUAUACAAUCUACUGUCUUUCCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038806
          miRBase ID: pbv-let-7f-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUCC
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-let-7a-1
          Stem-loop Accession # MI0039205
          Stem-loop Sequence
          UGAGGUAGUAGGUUGUAUAGUUUUAGGGUCACACCCACCACUGGGAGAUAACUAUACAAUCUACUGUCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048076
          miRBase ID: ocu-let-7a-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUCC
          Chromosome Location: NA

          Stem-loop ID ocu-let-7a-3
          Stem-loop Accession # MI0039207
          Stem-loop Sequence
          UGAGGUAGUAGGUUGUAUAGUUUGGGGCUCCGCCCUGCUAUGGGAUAACUAUACAAUCUACUGUCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048076
          miRBase ID: ocu-let-7a-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUCC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-let-7a-3
          Stem-loop Accession # MI0007572
          Stem-loop Sequence
          GGGUGAGGUAGUAGGUUGUAUAGUUUGGGGCUCUGCCCUGCUAUGGGAUAACUAUACAAUCUACUGUCUUUCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026789
          miRBase ID: mml-let-7a-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUCC
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-let-7e-1
          Stem-loop Accession # MI0026756
          Stem-loop Sequence
          UGAGGUAGUAGAUUGAAUAGUUGUGGGGUAUAUGUCCUCCCUCUGACAUAACUAUACAAUCUACUGUCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032706
          miRBase ID: ssa-let-7e-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUCC
          Chromosome Location: NA

          Stem-loop ID ssa-let-7e-2
          Stem-loop Accession # MI0026757
          Stem-loop Sequence
          UGAGGUAGUAGAUUGAAUAGUUGUGGGGUAUAUGUCCUCCCUCUGAGAUAACUAUACAAUCUACUGUCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032706
          miRBase ID: ssa-let-7e-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUCC
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-let-7a-1
          Stem-loop Accession # MI0013722
          Stem-loop Sequence
          UCCUUUGGGGUGAGGUAGUAGGUUGUAUAGUUUUAGGGCUAUGCCCUGCCUGUCAGAUAACUAUACAAUCUACUGUCUUUCCUGAAGUGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031085
          miRBase ID: tgu-let-7a-1-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUCC
          Chromosome Location: NA

          Stem-loop ID tgu-let-7e
          Stem-loop Accession # MI0013721
          Stem-loop Sequence
          UCCUUGAGGCUGAGGUAGUAGAUUGAAUAGUUGUGGAGUCUGGUCCUCCCUCUGAGCUAACUAUACAAUCUACUGUCUUUCCUAAGGAGACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026982
          miRBase ID: tgu-let-7e-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUCC
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-let-7a
          Stem-loop Accession # MI0031142
          Stem-loop Sequence
          UAAGGUAGUAGAUUGUAUAGUUCUAGGGUCACACCCAUCAUUGGAAGAUCACUCUAUACAAUCUACUGUCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036484
          miRBase ID: tch-let-7a-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUCC
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-let-7e
          Stem-loop Accession # MI0038222
          Stem-loop Sequence
          UGAGGUAGUAGGUUGUUUAGUUGUGGGUUGCACCCCGACAUUUGGAGAACUAUACAAUCUACUGUCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046402
          miRBase ID: xla-let-7e-3p
          Mature miRNA Sequence: CUAUACAAUCUACUGUCUUUCC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465223_pri
          TaqMan™ Pri-miRNA Assay : Rn03465703_pri
          TaqMan™ Pri-miRNA Assay : Mm03306770_pri
          TaqMan™ Pri-miRNA Assay : Mm03306744_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12649
          Ambion® Pre-miR™ miRNA Precursor : PM12649
          TaqMan™ MicroRNA Assay : 002478
          mirVana® miRNA inhibitor : MH12649
          mirVana® miRNA mimic : MC12649
          TaqMan™ Advanced miRNA Assay : rno481486_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline