Search
Search

| Assay Name: | mmu-miR-153-5p |
|---|---|
| Stem-loop Accession Number: | MI0000175 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GUCAUUUUUGUGACGUUGCAGCU |
| Species: | Mouse |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000050, mir-153 |
Mature miRNA Sequence: | GUCAUUUUUGUGACGUUGCAGCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-153-5p | MIMAT0016992 |