Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › rno478306_mir

          Specificity testing was performed using rat anti-targets.

          Assay Name: rno-miR-193a-3p
          Stem-loop Accession Number: MI0000487
          miRBase Version: v22.1
          Chromosome Location: Chr.17: 31559996 - 31560083 [+] on Build GRCh38
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Species: Human, Mouse, Rat, Astatotilapia burtoni, Bovine, Chicken, Cricetulus griseus, Cyprinus carpio, Dasypus novemcinctus, Eptesicus fuscus, Fugu rubripes, Guinea pig, Horse, Metriaclima zebra, Neolamprologus brichardi, Ophiophagus hannah, Oreochromis niloticus, Otolemur garnettii, Pan paniscus, Pan troglodytes, Papio hamadryas, Pig, Pongo pygmaeus, Pteropus alecto, Python bivittatus, Rabbit, Rhesus monkey, Tetraodon nigroviridis, Tupaia chinensis, Zebrafish
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID rno478306_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000082, mir-193

          Mature miRNA Sequence:

          AACUGGCCUACAAAGUCCCAGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-193a-3p MIMAT0000459
          Mouse mmu-miR-193a-3p MIMAT0000223
          Rat rno-miR-193a-3p MIMAT0000868
          Astatotilapia burtoni abu-miR-193-3p MIMAT0042161
          Bovine bta-miR-193a-3p MIMAT0003795
          Chicken gga-miR-193a-3p MIMAT0007740
          View More Rows
          Cricetulus griseus cgr-miR-193a MIMAT0041152
          Cyprinus carpio ccr-miR-193a MIMAT0026251
          Dasypus novemcinctus dno-miR-193a-3p MIMAT0047722
          Eptesicus fuscus efu-miR-193a MIMAT0035142
          Fugu rubripes fru-miR-193 MIMAT0002969
          Guinea pig cpo-miR-193a-3p MIMAT0047071
          Horse eca-miR-193a-3p MIMAT0013026
          Metriaclima zebra mze-miR-193 MIMAT0042391
          Neolamprologus brichardi nbr-miR-193 MIMAT0042573
          Ophiophagus hannah oha-miR-193-3p MIMAT0036780
          Oreochromis niloticus oni-miR-193 MIMAT0042763
          Otolemur garnettii oga-miR-193a MIMAT0049716
          Pan paniscus ppa-miR-193a MIMAT0049094
          Pan troglodytes ptr-miR-193a MIMAT0008058
          Papio hamadryas pha-miR-193a MIMAT0049518
          Pig ssc-miR-193a-3p MIMAT0013895
          Pongo pygmaeus ppy-miR-193a-3p MIMAT0015788
          Pteropus alecto pal-miR-193a-3p MIMAT0039932
          Python bivittatus pbv-miR-193a-3p MIMAT0038990
          Rabbit ocu-miR-193a-3p MIMAT0048304
          Rhesus monkey mml-miR-193a-3p MIMAT0006226
          Tetraodon nigroviridis tni-miR-193 MIMAT0002970
          Tupaia chinensis tch-miR-193a-3p MIMAT0036584
          Zebrafish dre-miR-193a-3p MIMAT0001856

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-193a
          Stem-loop Accession # MI0000487
          Stem-loop Sequence
          CGAGGAUGGGAGCUGAGGGCUGGGUCUUUGCGGGCGAGAUGAGGGUGUCGGAUCAACUGGCCUACAAAGUCCCAGUUCUCGGCCCCCG
          Chromosome Location Chr. 17 - 31559996 - 31560083 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000459
          miRBase ID: hsa-miR-193a-3p
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: Chr. 17 - 31559996 - 31560083 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-193a
          Stem-loop Accession # MI0000235
          Stem-loop Sequence
          GAGAGCUGGGUCUUUGCGGGCAAGAUGAGAGUGUCAGUUCAACUGGCCUACAAAGUCCCAGUCCUC
          Chromosome Location Chr. 11 - 79711969 - 79712034 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000223
          miRBase ID: mmu-miR-193a-3p
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: Chr. 11 - 79711969 - 79712034 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-193a
          Stem-loop Accession # MI0000936
          Stem-loop Sequence
          GCGGACGGGAGCUGAGAGCUGGGUCUUUGCGGGCAAGAUGAGGGUGUCAGUUCAACUGGCCUACAAAGUCCCAGUCCUCGGCUCCC
          Chromosome Location Chr. 10 - 67065896 - 67065981 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000868
          miRBase ID: rno-miR-193a-3p
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: Chr. 10 - 67065896 - 67065981 [+] on Build Rnor_6.0

          Astatotilapia burtoni

          Stem-loop ID abu-mir-193
          Stem-loop Accession # MI0033876
          Stem-loop Sequence
          UGGGUCUUUGCGGGCAAGGUGAGUCCUCAUUUCAUUCAACUGGCCUACAAAGUCCCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042161
          miRBase ID: abu-miR-193-3p
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-193a
          Stem-loop Accession # MI0005014
          Stem-loop Sequence
          UGGGAGCUGAGAGCUGGGUCUUUGCGGGCGAGAUGAAGGUGUCGGUUCAACUGGCCUACAAAGUCCCAGUCCUCGGCCCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003795
          miRBase ID: bta-miR-193a-3p
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-193a
          Stem-loop Accession # MI0007564
          Stem-loop Sequence
          AGCUGAGGGCUGGGUCUUUGCGGGCGAGAUGAGAGGUUCGUGUCUUCAACUGGCCUACAAAGUCCCAGUUCUCGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007740
          miRBase ID: gga-miR-193a-3p
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-193a
          Stem-loop Accession # MI0033081
          Stem-loop Sequence
          GAGAGCUGGGUCUUUGCGGGCAAGAUGAGGGUGUCAGUUCAACUGGCCUACAAAGUCCCAGUCCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041152
          miRBase ID: cgr-miR-193a
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: NA

          Cyprinus carpio

          Stem-loop ID ccr-mir-193a
          Stem-loop Accession # MI0023355
          Stem-loop Sequence
          AAAGACAUUAAUGUGUUAGAGGUUGGGUCUUUGCGGGCAAGGUGAGUAGUUAAAUUUACUCUCAACUGGCCUACAAAGUCCCAGUUUCUGGCUCAUGUAAUCAUCCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026251
          miRBase ID: ccr-miR-193a
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-193a
          Stem-loop Accession # MI0039024
          Stem-loop Sequence
          UGGGUCUUUGCGGGCGAGAUGAGGGUGUCGGUUCAACUGGCCUACAAAGUCCCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047722
          miRBase ID: dno-miR-193a-3p
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: NA

          Eptesicus fuscus

          Stem-loop ID efu-mir-193a
          Stem-loop Accession # MI0028845
          Stem-loop Sequence
          AGUUGAGGGCUGGGUCUUUGCGGGCGAGAUGAGGGUGUCGGUUCAACUGGCCUACAAAGUCCCAGUCCUCGGCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035142
          miRBase ID: efu-miR-193a
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: NA

          Fugu rubripes

          Stem-loop ID fru-mir-193
          Stem-loop Accession # MI0003285
          Stem-loop Sequence
          UGUGUCAGAAGCUGGGUCUUUGCGGGCAAGGUGAGUCCUCAGUUUGUUCAACUGGCCUACAAAGUCCCAGUCUCUGGCUCACGUAACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002969
          miRBase ID: fru-miR-193
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-193a
          Stem-loop Accession # MI0038681
          Stem-loop Sequence
          UGGGUCUUUGCGGGCGAGAUGAGGGUGUCGGUUCAACUGGCCUACAAAGUCCCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047071
          miRBase ID: cpo-miR-193a-3p
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-193a
          Stem-loop Accession # MI0012773
          Stem-loop Sequence
          GGGAGCUGAGGGCUGGGUCUUUGCGGGCGAGAUGAGGGUGUCAGUUCAACUGGCCUACAAAGUCCCAGUCUUCGGCCCCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013026
          miRBase ID: eca-miR-193a-3p
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: NA

          Metriaclima zebra

          Stem-loop ID mze-mir-193
          Stem-loop Accession # MI0034175
          Stem-loop Sequence
          UGGGUCUUUGCGGGCAAGGUGAGUCCUCAUUUCAUUCAACUGGCCUACAAAGUCCCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042391
          miRBase ID: mze-miR-193
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: NA

          Neolamprologus brichardi

          Stem-loop ID nbr-mir-193
          Stem-loop Accession # MI0034431
          Stem-loop Sequence
          UGGGUCUUUGCGGGCAAGGUGAGUCCUCAUUUCAUUCAACUGGCCUACAAAGUCCCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042573
          miRBase ID: nbr-miR-193
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-193
          Stem-loop Accession # MI0031387
          Stem-loop Sequence
          UGGGGCUGGGUCUUUGCGGGCGAGAUGAGAAGUUUCUGCCUUCAACUGGCCUACAAAGUCCCAGUUCUCGGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036780
          miRBase ID: oha-miR-193-3p
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: NA

          Oreochromis niloticus

          Stem-loop ID oni-mir-193
          Stem-loop Accession # MI0034686
          Stem-loop Sequence
          UGGGUCUUUGCGGGCAAGGUGAGUCCUCAUUUCAUUCAACUGGCCUACAAAGUCCCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042763
          miRBase ID: oni-miR-193
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-193a
          Stem-loop Accession # MI0040504
          Stem-loop Sequence
          UGGGUCUUUGCGGGCGAGAUGAGGAUGUCGGAUCAACUGGCCUACAAAGUCCCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049716
          miRBase ID: oga-miR-193a
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-193a
          Stem-loop Accession # MI0039804
          Stem-loop Sequence
          UGGGUCUUUGCGGGCGAGAUGAGGGUGUCGGAUCAACUGGCCUACAAAGUCCCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049094
          miRBase ID: ppa-miR-193a
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-193a
          Stem-loop Accession # MI0008566
          Stem-loop Sequence
          CGAGGAUGGGAGCUGAGGGCUGGGUCUUUGCGGGCGAGAUGAGGGUGUCGGAUCAACUGGCCUACAAAGUCCCAGUUCUCGGCCCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008058
          miRBase ID: ptr-miR-193a
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: NA

          Papio hamadryas

          Stem-loop ID pha-mir-193a
          Stem-loop Accession # MI0040283
          Stem-loop Sequence
          UGGGUCUUUGCGGGCGAGAUGAGGGUGUCGGAUCAACUGGCCUACAAAGUCCCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049518
          miRBase ID: pha-miR-193a
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-193a
          Stem-loop Accession # MI0013113
          Stem-loop Sequence
          GCGGGAGCUGAGGGCUGGGUCUUUGCGGGCGAGAUGAGGGUGUUGGUUCAACUGGCCUACAAAGUCCCAGUCCUCGGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013895
          miRBase ID: ssc-miR-193a-3p
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-193a
          Stem-loop Accession # MI0014855
          Stem-loop Sequence
          CGAGGAUGGGAGCUGAGGGCUGGGUCUUUGCGGGCGAGAUGAAGGUGUCGGAUCAACUGGCCUACAAAGUCCCAGUCCUCGGCCCCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015788
          miRBase ID: ppy-miR-193a-3p
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-193a
          Stem-loop Accession # MI0032406
          Stem-loop Sequence
          UGGGUCUUUGCGGGCGAGAUGAGGGUGUCGAUUCAACUGGCCUACAAAGUCCCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039932
          miRBase ID: pal-miR-193a-3p
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-193a
          Stem-loop Accession # MI0030265
          Stem-loop Sequence
          GAGCAGCUGGGAGUUUGGGGCUGGGUCUUUGCGGGCGAGAUGAGAAGUUUCUGCCUUCAACUGGCCUACAAAGUCCCAGUUCUCGGCUCUUAACUUUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038990
          miRBase ID: pbv-miR-193a-3p
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-193a
          Stem-loop Accession # MI0039330
          Stem-loop Sequence
          UGGGUCUUUGCGGGCGAGAUGAGGGUGUCGGUUCAACUGGCCUACAAAGUCCCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048304
          miRBase ID: ocu-miR-193a-3p
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-193a
          Stem-loop Accession # MI0007655
          Stem-loop Sequence
          GGAUGGGAGCUGAGGGCUGGGUCUUUGCGGGCGAGAUGAGGGUGUCGGAUCAACUGGCCUACAAAGUCCCAGUCCUCGGCCCCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006226
          miRBase ID: mml-miR-193a-3p
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: NA

          Tetraodon nigroviridis

          Stem-loop ID tni-mir-193
          Stem-loop Accession # MI0003286
          Stem-loop Sequence
          CAGAAGCUGGGUCUUUGCGGGCAAGGUGAGUCCUCAGUUUGUUCAACUGGCCUACAAAGUCCCAGUGUCUGGCUCACGUGACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002970
          miRBase ID: tni-miR-193
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-193a-1
          Stem-loop Accession # MI0031248
          Stem-loop Sequence
          UGGGUCUUUGCGGGCAAGAUGAGGGUGUUGGUUCAACUGGCCUACAAAGUCCCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036584
          miRBase ID: tch-miR-193a-3p
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: NA

          Stem-loop ID tch-mir-193a-2
          Stem-loop Accession # MI0031268
          Stem-loop Sequence
          UGGGUCUUUGCGGGCAAGAUGAGGGUGUUGGUUCAACUGGCCUACAAAGUCCCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036584
          miRBase ID: tch-miR-193a-3p
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-193a-1
          Stem-loop Accession # MI0002029
          Stem-loop Sequence
          GACAUUAAUGUGUUAGAGGUUGGGUCUUUGCGGGCAAGGUGAGUAGUUAAAUUUACUCUCAACUGGCCUACAAAGUCCCAGUUUCUGGCUCAUGUUAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001856
          miRBase ID: dre-miR-193a-3p
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: NA

          Stem-loop ID dre-mir-193a-2
          Stem-loop Accession # MI0002030
          Stem-loop Sequence
          GAGAGUGUGUCAGAGGCUGGGUCUUUGCGGGCAAGGUGAGUUUUCCUUUCAUUCAACUGGCCUACAAAGUCCCAGUUUUCGGCCCAUGUGACCAUCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001856
          miRBase ID: dre-miR-193a-3p
          Mature miRNA Sequence: AACUGGCCUACAAAGUCCCAGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303307_pri
          TaqMan™ Pri-miRNA Assay : Rn03465100_pri
          TaqMan™ Pri-miRNA Assay : Mm03306486_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11123
          Ambion® Pre-miR™ miRNA Precursor : PM11123
          TaqMan™ MicroRNA Assay : 002250
          mirVana® miRNA inhibitor : MH11123
          mirVana® miRNA mimic : MC11123
          TaqMan™ Advanced miRNA Assay : mmu478306_mir
          TaqMan™ Advanced miRNA Assay : 478306_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline