Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › rno478318_mir

          Specificity testing was performed using rat anti-targets.

          Assay Name: rno-miR-212-3p
          Stem-loop Accession Number: MI0000696
          miRBase Version: v22.1
          Chromosome Location: Chr.11: 75173388 - 75173478 [+] on Build GRCm38
          Mature miRNA Sequence: UAACAGUCUCCAGUCACGGCCA
          Species: Mouse, Rat, Guinea pig, Pteropus alecto, Rabbit
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID rno478318_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000065, mir-132

          Mature miRNA Sequence:

          UAACAGUCUCCAGUCACGGCCA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-212-3p MIMAT0000659
          Rat rno-miR-212-3p MIMAT0000883
          Guinea pig cpo-miR-212-3p MIMAT0047114
          Pteropus alecto pal-miR-212-3p MIMAT0039980
          Rabbit ocu-miR-212-3p MIMAT0048336

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-212
          Stem-loop Accession # MI0000696
          Stem-loop Sequence
          GGGCAGCGCGCCGGCACCUUGGCUCUAGACUGCUUACUGCCCGGGCCGCCUUCAGUAACAGUCUCCAGUCACGGCCACCGACGCCUGGCCC
          Chromosome Location Chr. 11 - 75173388 - 75173478 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000659
          miRBase ID: mmu-miR-212-3p
          Mature miRNA Sequence: UAACAGUCUCCAGUCACGGCCA
          Chromosome Location: Chr. 11 - 75173388 - 75173478 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-212
          Stem-loop Accession # MI0000952
          Stem-loop Sequence
          CGGGAUAUCCCCGCCCGGGCAGCGCGCCGGCACCUUGGCUCUAGACUGCUUACUGCCCGGGCCGCCCUCAGUAACAGUCUCCAGUCACGGCCACCGACGCCUGGCCCCGCC
          Chromosome Location Chr. 10 - 62014702 - 62014812 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000883
          miRBase ID: rno-miR-212-3p
          Mature miRNA Sequence: UAACAGUCUCCAGUCACGGCCA
          Chromosome Location: Chr. 10 - 62014702 - 62014812 [+] on Build Rnor_6.0

          Guinea pig

          Stem-loop ID cpo-mir-212
          Stem-loop Accession # MI0038704
          Stem-loop Sequence
          ACCUUGGCUCUAGACUGCUUACUGCCCGGGCCGCCCUCAGUAACAGUCUCCAGUCACGGCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047114
          miRBase ID: cpo-miR-212-3p
          Mature miRNA Sequence: UAACAGUCUCCAGUCACGGCCA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-212
          Stem-loop Accession # MI0032431
          Stem-loop Sequence
          ACCUUGGCUCUAGACUGCUUACUGCCCGGGCCGCCCUCAGUAACAGUCUCCAGUCACGGCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039980
          miRBase ID: pal-miR-212-3p
          Mature miRNA Sequence: UAACAGUCUCCAGUCACGGCCA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-212
          Stem-loop Accession # MI0039347
          Stem-loop Sequence
          ACCUUGGCUCUAGACUGCUUACUGCCCGGGCCGCCCUCAGUAACAGUCUCCAGUCACGGCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048336
          miRBase ID: ocu-miR-212-3p
          Mature miRNA Sequence: UAACAGUCUCCAGUCACGGCCA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307114_pri
          TaqMan™ Pri-miRNA Assay : Rn03465057_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12534
          Ambion® Pre-miR™ miRNA Precursor : PM12534
          TaqMan™ MicroRNA Assay : 002551
          mirVana® miRNA inhibitor : MH12534
          mirVana® miRNA mimic : MC12534
          TaqMan™ Advanced miRNA Assay : mmu478318_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline