Search
Search

| Assay Name: | rno-miR-125a-5p |
|---|---|
| Stem-loop Accession Number: | MI0000469 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCCCUGAGACCCUUUAACCUGUGA |
| Species: | Human, Mouse, Rat, Cricetulus griseus, Guinea pig, Horse, Otolemur garnettii, Pongo pygmaeus, Rhesus monkey, Saimiri boliviensis |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000033, mir-10 |
Mature miRNA Sequence: | UCCCUGAGACCCUUUAACCUGUGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-125a-5p | MIMAT0000443 |
| Mouse | mmu-miR-125a-5p | MIMAT0000135 |
| Rat | rno-miR-125a-5p | MIMAT0000829 |
| Cricetulus griseus | cgr-miR-125a-5p | MIMAT0023741 |
| Guinea pig | cpo-miR-125a-5p | MIMAT0046965 |
| Horse | eca-miR-125a-5p | MIMAT0013008 |
| View More Rows | ||
| Otolemur garnettii | oga-miR-125a | MIMAT0049651 |
| Pongo pygmaeus | ppy-miR-125a-5p | MIMAT0015748 |
| Rhesus monkey | mml-miR-125a-5p | MIMAT0006181 |
| Saimiri boliviensis | sbo-miR-125a | MIMAT0049472 |