Search
Search

| Assay Name: | rno-miR-138-1-3p |
|---|---|
| Stem-loop Accession Number: | MI0000722 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CGGCUACUUCACAACACCAGGG |
| Species: | Mouse, Rat, Anolis carolinensis, Ophiophagus hannah |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000075, mir-138 |
Mature miRNA Sequence: | CGGCUACUUCACAACACCAGGG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-138-1-3p | MIMAT0004668 |
| Rat | rno-miR-138-1-3p | MIMAT0004734 |
| Anolis carolinensis | aca-miR-138-2-3p | MIMAT0021759 |
| Ophiophagus hannah | oha-miR-138-1-3p | MIMAT0036711 |