Search
Search

| Assay Name: | rno-miR-188-5p |
|---|---|
| Stem-loop Accession Number: | MI0000484 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAUCCCUUGCAUGGUGGAGGG |
| Species: | Human, Mouse, Rat, Dasypus novemcinctus, Goat, Guinea pig, Horse, Pteropus alecto, Rabbit |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000113, mir-188 |
Mature miRNA Sequence: | CAUCCCUUGCAUGGUGGAGGG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-188-5p | MIMAT0000457 |
| Mouse | mmu-miR-188-5p | MIMAT0000217 |
| Rat | rno-miR-188-5p | MIMAT0005301 |
| Dasypus novemcinctus | dno-miR-188-5p | MIMAT0047711 |
| Goat | chi-miR-188-5p | MIMAT0036013 |
| Guinea pig | cpo-miR-188-5p | MIMAT0047060 |
| View More Rows | ||
| Horse | eca-miR-188-5p | MIMAT0013198 |
| Pteropus alecto | pal-miR-188-5p | MIMAT0040345 |
| Rabbit | ocu-miR-188-5p | MIMAT0048297 |