Search
Search

| Assay Name: | rno-miR-3587 |
|---|---|
| Stem-loop Accession Number: | MI0015463 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AACACUGAUUUCAAAUGGUGC |
| Species: | Rat |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Mature miRNA Sequence: | AACACUGAUUUCAAAUGGUGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-3587 | MIMAT0017883 |