Search
Search

| Assay Name: | rno-miR-375-5p |
|---|---|
| Stem-loop Accession Number: | MI0000783 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GCGACGAGCCCCUCGCACAAACC |
| Species: | Human, Rat, Dasypus novemcinctus, Guinea pig, Pteropus alecto, Rabbit |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000114, mir-375 |
Mature miRNA Sequence: | GCGACGAGCCCCUCGCACAAACC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-375-5p | MIMAT0037313 |
| Rat | rno-miR-375-5p | MIMAT0017301 |
| Dasypus novemcinctus | dno-miR-375-5p | MIMAT0047848 |
| Guinea pig | cpo-miR-375-5p | MIMAT0047207 |
| Pteropus alecto | pal-miR-375-5p | MIMAT0040120 |
| Rabbit | ocu-miR-375-5p | MIMAT0048410 |