Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › rno481278_mir

          Specificity testing was performed using rat anti-targets.

          Assay Name: rno-miR-92b-5p
          Stem-loop Accession Number: MI0006167
          miRBase Version: v22.1
          Chromosome Location: Chr.2: 188541034 - 188541116 [-] on Build Rnor_6.0
          Mature miRNA Sequence: AGGGACGGGACGCGGUGCAGUGUU
          Species: Rat, Cricetulus griseus, Guinea pig, Pig, Rhesus monkey
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID rno481278_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000013, mir-25

          Mature miRNA Sequence:

          AGGGACGGGACGCGGUGCAGUGUU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Rat rno-miR-92b-5p MIMAT0017319
          Cricetulus griseus cgr-miR-92b-5p MIMAT0024011
          Guinea pig cpo-miR-92b-5p MIMAT0046931
          Pig ssc-miR-92b-5p MIMAT0017377
          Rhesus monkey mml-miR-92b-5p MIMAT0026810

          Stem-loop Details



          Rat

          Stem-loop ID rno-mir-92b
          Stem-loop Accession # MI0006167
          Stem-loop Sequence
          GGUGGGCAGGAGGGACGGGACGCGGUGCAGUGUUGUUCUUUCCCCUGCCAAUAUUGCACUCGUCCCGGCCUCCGGCCCCCUCG
          Chromosome Location Chr. 2 - 188541034 - 188541116 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017319
          miRBase ID: rno-miR-92b-5p
          Mature miRNA Sequence: AGGGACGGGACGCGGUGCAGUGUU
          Chromosome Location: Chr. 2 - 188541034 - 188541116 [-] on Build Rnor_6.0

          Cricetulus griseus

          Stem-loop ID cgr-mir-92b
          Stem-loop Accession # MI0020561
          Stem-loop Sequence
          GGCGGGCGGGAGGGACGGGACGCGGUGCAGUGUUGUUCUUUCCCCCGCCAAUAUUGCACUCGUCCCGGCCUCCGGCCCCCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024011
          miRBase ID: cgr-miR-92b-5p
          Mature miRNA Sequence: AGGGACGGGACGCGGUGCAGUGUU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-92b
          Stem-loop Accession # MI0038605
          Stem-loop Sequence
          AGGGACGGGACGCGGUGCAGUGUUGUUCUUUCCCCCACCAAUAUUGCACUCGUCCCGGCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046931
          miRBase ID: cpo-miR-92b-5p
          Mature miRNA Sequence: AGGGACGGGACGCGGUGCAGUGUU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-92b
          Stem-loop Accession # MI0013126
          Stem-loop Sequence
          GCGGGCGGGAGGGACGGGACGCGGUGCAGUGUUGUUCUUUCCCCUGCCAAUAUUGCACUCGUCCCGGCCUCCGGCCCCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0017377
          miRBase ID: ssc-miR-92b-5p
          Mature miRNA Sequence: AGGGACGGGACGCGGUGCAGUGUU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-92b
          Stem-loop Accession # MI0007607
          Stem-loop Sequence
          CGGGCCCCGGGCGGGCGGGAGGGACGGGACGCGGUGCAGUGUUGUUCUUUCCCCCGCCAAUAUUGCACUCGUCCCGGCCUCCGGCCCCCCCGGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026810
          miRBase ID: mml-miR-92b-5p
          Mature miRNA Sequence: AGGGACGGGACGCGGUGCAGUGUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03464732_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM20550
          Ambion® Pre-miR™ miRNA Precursor : PM20550
          TaqMan™ MicroRNA Assay : 463385_mat
          mirVana® miRNA inhibitor : MH20550
          mirVana® miRNA mimic : MC20550

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline