Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › rno481279_mir

          Specificity testing was performed using rat anti-targets.

          Assay Name: rno-miR-93-3p
          Stem-loop Accession Number: MI0000880
          miRBase Version: v22.1
          Chromosome Location: Chr.12: 19307543 - 19307629 [-] on Build Rnor_6.0
          Mature miRNA Sequence: ACUGCUGAGCUAGCACUUCCCGA
          Species: Rat, Cricetulus griseus, Dasypus novemcinctus, Guinea pig, Rabbit
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID rno481279_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000001, mir-17

          Mature miRNA Sequence:

          ACUGCUGAGCUAGCACUUCCCGA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Rat rno-miR-93-3p MIMAT0017109
          Cricetulus griseus cgr-miR-93-3p MIMAT0024014
          Dasypus novemcinctus dno-miR-93-3p MIMAT0047587
          Guinea pig cpo-miR-93-3p MIMAT0046934
          Rabbit ocu-miR-93-3p MIMAT0048177

          Stem-loop Details



          Rat

          Stem-loop ID rno-mir-93
          Stem-loop Accession # MI0000880
          Stem-loop Sequence
          AGUCAUGGGGGCUCCAAAGUGCUGUUCGUGCAGGUAGUGCAUUGCCUGACCUACUGCUGAGCUAGCACUUCCCGAGCCCCCAGGACA
          Chromosome Location Chr. 12 - 19307543 - 19307629 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017109
          miRBase ID: rno-miR-93-3p
          Mature miRNA Sequence: ACUGCUGAGCUAGCACUUCCCGA
          Chromosome Location: Chr. 12 - 19307543 - 19307629 [-] on Build Rnor_6.0

          Cricetulus griseus

          Stem-loop ID cgr-mir-93
          Stem-loop Accession # MI0020562
          Stem-loop Sequence
          AUGGGGGCUCCAAAGUGCUGUUCGUGCAGGUAGUGUAAUUACCUGACCUACUGCUGAGCUAGCACUUCCCGAGCCCCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024014
          miRBase ID: cgr-miR-93-3p
          Mature miRNA Sequence: ACUGCUGAGCUAGCACUUCCCGA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-93
          Stem-loop Accession # MI0038950
          Stem-loop Sequence
          CAAAGUGCUGUUCGUGCAGGUAGUGUGAUUACCUGACCUACUGCUGAGCUAGCACUUCCCGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047587
          miRBase ID: dno-miR-93-3p
          Mature miRNA Sequence: ACUGCUGAGCUAGCACUUCCCGA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-93
          Stem-loop Accession # MI0038606
          Stem-loop Sequence
          CAAAGUGCUGUUCGUGCAGGUAGUGUGAUUACCUGACCUACUGCUGAGCUAGCACUUCCCGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046934
          miRBase ID: cpo-miR-93-3p
          Mature miRNA Sequence: ACUGCUGAGCUAGCACUUCCCGA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-93
          Stem-loop Accession # MI0039261
          Stem-loop Sequence
          CAAAGUGCUGUUCGUGCAGGUAGUGUGAUUACCUGACCUACUGCUGAGCUAGCACUUCCCGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048177
          miRBase ID: ocu-miR-93-3p
          Mature miRNA Sequence: ACUGCUGAGCUAGCACUUCCCGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465019_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM19859
          Ambion® Pre-miR™ miRNA Precursor : PM19859
          TaqMan™ MicroRNA Assay : 462039_mat
          mirVana® miRNA inhibitor : MH19859
          mirVana® miRNA mimic : MC19859

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline