Search
Search

| Assay Name: | rno-miR-879-5p |
|---|---|
| Stem-loop Accession Number: | MI0005472 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGAGGCUUAUAGCUCUAAGCC |
| Species: | Mouse, Rat |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000421, mir-879 |
Mature miRNA Sequence: | AGAGGCUUAUAGCUCUAAGCC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-879-5p | MIMAT0004842 |
| Rat | rno-miR-879-5p | MIMAT0005287 |