Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › rno481491_mir

          Specificity testing was performed using rat anti-targets.

          Assay Name: rno-miR-133a-3p
          Stem-loop Accession Number: MI0000450
          miRBase Version: v22.1
          Chromosome Location: Chr.18: 21825698 - 21825785 [-] on Build GRCh38
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUG
          Species: Human, Mouse, Rat, Alligator mississippiensis, Anolis carolinensis, Bovine, Chrysemys picta, Fugu rubripes, Horse, Ophiophagus hannah, Pan troglodytes, Pongo pygmaeus, Rhesus monkey, Tetraodon nigroviridis, Zebrafish
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID rno481491_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000029, mir-133

          Mature miRNA Sequence:

          UUUGGUCCCCUUCAACCAGCUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-133a-3p MIMAT0000427
          Mouse mmu-miR-133a-3p MIMAT0000145
          Rat rno-miR-133a-3p MIMAT0000839
          Alligator mississippiensis ami-miR-133a-3p MIMAT0038147
          Anolis carolinensis aca-miR-133a MIMAT0021749
          Anolis carolinensis aca-miR-133a-3p MIMAT0037566
          View More Rows
          Bovine bta-miR-133a MIMAT0009225
          Chrysemys picta cpi-miR-133c-3p MIMAT0037753
          Fugu rubripes fru-miR-133 MIMAT0003057
          Horse eca-miR-133a MIMAT0012997
          Ophiophagus hannah oha-miR-133a-3p MIMAT0036699
          Pan troglodytes ptr-miR-133a MIMAT0002243
          Pongo pygmaeus ppy-miR-133c MIMAT0016973
          Rhesus monkey mml-miR-133c-3p MIMAT0006777
          Tetraodon nigroviridis tni-miR-133 MIMAT0003058
          Zebrafish dre-miR-133a-3p MIMAT0001830

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-133a-1
          Stem-loop Accession # MI0000450
          Stem-loop Sequence
          ACAAUGCUUUGCUAGAGCUGGUAAAAUGGAACCAAAUCGCCUCUUCAAUGGAUUUGGUCCCCUUCAACCAGCUGUAGCUAUGCAUUGA
          Chromosome Location Chr. 18 - 21825698 - 21825785 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000427
          miRBase ID: hsa-miR-133a-3p
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUG
          Chromosome Location: Chr. 18 - 21825698 - 21825785 [-] on Build GRCh38

          Stem-loop ID hsa-mir-133a-2
          Stem-loop Accession # MI0000451
          Stem-loop Sequence
          GGGAGCCAAAUGCUUUGCUAGAGCUGGUAAAAUGGAACCAAAUCGACUGUCCAAUGGAUUUGGUCCCCUUCAACCAGCUGUAGCUGUGCAUUGAUGGCGCCG
          Chromosome Location Chr. 20 - 62564912 - 62565013 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000427
          miRBase ID: hsa-miR-133a-3p
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUG
          Chromosome Location: Chr. 18 - 21825698 - 21825785 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-133a-1
          Stem-loop Accession # MI0000159
          Stem-loop Sequence
          GCUAAAGCUGGUAAAAUGGAACCAAAUCGCCUCUUCAAUGGAUUUGGUCCCCUUCAACCAGCUGUAGC
          Chromosome Location Chr. 18 - 10782909 - 10782976 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000145
          miRBase ID: mmu-miR-133a-3p
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUG
          Chromosome Location: Chr. 18 - 10782909 - 10782976 [-] on Build GRCm38

          Stem-loop ID mmu-mir-133a-2
          Stem-loop Accession # MI0000820
          Stem-loop Sequence
          AGAAGCCAAAUGCUUUGCUGAAGCUGGUAAAAUGGAACCAAAUCAGCUGUUGGAUGGAUUUGGUCCCCUUCAACCAGCUGUAGCUGCGCAUUGAUCACGCCGCA
          Chromosome Location Chr. 2 - 180398379 - 180398482 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000145
          miRBase ID: mmu-miR-133a-3p
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUG
          Chromosome Location: Chr. 18 - 10782909 - 10782976 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-133a
          Stem-loop Accession # MI0000906
          Stem-loop Sequence
          CAAUGCUUUGCUAAAGCUGGUAAAAUGGAACCAAAUCGCCUCUUCAAUGGAUUUGGUCCCCUUCAACCAGCUGUAGCUAUGCAUUGA
          Chromosome Location Chr. 18 - 2052478 - 2052564 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000839
          miRBase ID: rno-miR-133a-3p
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUG
          Chromosome Location: Chr. 18 - 2052478 - 2052564 [-] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-133a-1
          Stem-loop Accession # MI0029733
          Stem-loop Sequence
          CCAAUGCUUUGCUAAAGCUGGUAAAAUGGAACCAAAUCACCUGUUCAAUGGAUUUGGUCCCCUUCAACCAGCUGUAGCUAUGCAUUGAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038147
          miRBase ID: ami-miR-133a-3p
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUG
          Chromosome Location: NA

          Stem-loop ID ami-mir-133a-2
          Stem-loop Accession # MI0029734
          Stem-loop Sequence
          CCAAAUGCUUUGCUAAAGCUGGUAAAAUGGAACCAAAUCAACUGUUCAAUGGAUUUGGUCCCCUUCAACCAGCUGUAGCUGUGCAUUGAUCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038147
          miRBase ID: ami-miR-133a-3p
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUG
          Chromosome Location: NA

          Stem-loop ID ami-mir-133a-3
          Stem-loop Accession # MI0029735
          Stem-loop Sequence
          AGUGUGUUCCUGGGGCUGGUAAAAAGGAACCAGAUCAACUUGGAACUGGAUUUGGUCCCCUUCAACCAGCUGUAGUGGCACAUAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038147
          miRBase ID: ami-miR-133a-3p
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUG
          Chromosome Location: NA

          Anolis carolinensis

          Stem-loop ID aca-mir-133a-1
          Stem-loop Accession # MI0018739
          Stem-loop Sequence
          GAGGCCCAGAAUGCUUUGCUAAAGCUGGUAAAAUGGAACCAAAUCAACUGUUCAAUGGAUUUGGUCCCCUUCAACCAGCUGUAGCUGUGCAUUGAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021749
          miRBase ID: aca-miR-133a
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUG
          Chromosome Location: NA

          Stem-loop ID aca-mir-133a-2
          Stem-loop Accession # MI0018740
          Stem-loop Sequence
          CUUAAUGUCGAUGCUUUGCUAAAGCUGGUAAAAUGGAACCAAAUCACCUGUUAAAUGGAUUUGGUCCCCUUCAACCAGCUGUAGCUAUGUAUUGAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021749
          miRBase ID: aca-miR-133a
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUG
          Chromosome Location: NA

          Stem-loop ID aca-mir-133a-3
          Stem-loop Accession # MI0029353
          Stem-loop Sequence
          UGUGUCCCUAGGGCUGGUAAAAAGGAACCAGAUCGACUGGCAACUGGAUUUGGUCCCCUUCAACCAGCUGUGGUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037566
          miRBase ID: aca-miR-133a-3p
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUG
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-133a-1
          Stem-loop Accession # MI0009733
          Stem-loop Sequence
          UGGGACCGAAUGCUUUGCUAAAGCUGGUAAAAUGGAACCAAAUCAACUGUUCGAUGGAUUUGGUCCCCUUCAACCAGCUGUAGCUGCGCAUUGAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009225
          miRBase ID: bta-miR-133a
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUG
          Chromosome Location: NA

          Stem-loop ID bta-mir-133a-2
          Stem-loop Accession # MI0009732
          Stem-loop Sequence
          CAAUGCUUUGCUAGAGCUGGUAAAAUGGAACCAAAUCGCCUCUUCAAUGGAUUUGGUCCCCUUCAACCAGCUGUAGCUAUGCAUUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009225
          miRBase ID: bta-miR-133a
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUG
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-133c
          Stem-loop Accession # MI0029475
          Stem-loop Sequence
          CUGGAGAGAGUGUGUCCCUGGGGCUGGUAAAAAGGAACCAGAUCAACUUGGAACUGGAUUUGGUCCCCUUCAACCAGCUGCAGUGGCGCAUACGCCAGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037753
          miRBase ID: cpi-miR-133c-3p
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUG
          Chromosome Location: NA

          Fugu rubripes

          Stem-loop ID fru-mir-133
          Stem-loop Accession # MI0003397
          Stem-loop Sequence
          AAUGCUUUGCUAAAGCUGGUAAAAUGGAACCAAAUCACCUCUUGAAUGGAUUUGGUCCCCUUCAACCAGCUGUAGCUAUGCUUUGAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003057
          miRBase ID: fru-miR-133
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-133a
          Stem-loop Accession # MI0012749
          Stem-loop Sequence
          GCUAGAGCUGGUAAAAUGGAACCAAAUCGCCUCUUCAAUGGAUUUGGUCCCCUUCAACCAGCUGUAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012997
          miRBase ID: eca-miR-133a
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUG
          Chromosome Location: NA

          Stem-loop ID eca-mir-133a-2
          Stem-loop Accession # MI0012856
          Stem-loop Sequence
          GCUAAAGCUGGUAAAAUGGAACCAAAUCAACUGUUCAAUGGAUUUGGUCCCCUUCAACCAGCUGUAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012997
          miRBase ID: eca-miR-133a
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUG
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-133a-1
          Stem-loop Accession # MI0031341
          Stem-loop Sequence
          CUUUGCUUAAGCUGGUAAAAUGGAAUCAAAUCGCAUGUUUAAUGGAUUUGGUCCCCUUCAACCAGCUGUAGCUAUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036699
          miRBase ID: oha-miR-133a-3p
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUG
          Chromosome Location: NA

          Stem-loop ID oha-mir-133a-2
          Stem-loop Accession # MI0031342
          Stem-loop Sequence
          CUUUGCUAAAGCUGGUAAAAUGGAACCAAAUCAAGUGUUCAAUGGAUUUGGUCCCCUUCAACCAGCUGUAGCUGUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036699
          miRBase ID: oha-miR-133a-3p
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUG
          Chromosome Location: NA

          Stem-loop ID oha-mir-133a-3
          Stem-loop Accession # MI0031344
          Stem-loop Sequence
          UGUGCUCCUGGGGCUGGUAAAAAGGAACCAGAUCGACUGGCGACUGAUUUGGUCCCCUUCAACCAGCUGGGGGGGCACAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036699
          miRBase ID: oha-miR-133a-3p
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-133a-1
          Stem-loop Accession # MI0002532
          Stem-loop Sequence
          ACAAUGCUUUGCUAGAGCUGGUAAAAUGGAACCAAAUCGCCUCUUCAAUGGAUUUGGUCCCCUUCAACCAGCUGUAGCUAUGCAUUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002243
          miRBase ID: ptr-miR-133a
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUG
          Chromosome Location: NA

          Stem-loop ID ptr-mir-133a-2
          Stem-loop Accession # MI0008533
          Stem-loop Sequence
          GGAACCAAAUGCUUUGCUAGAGCUGGUAAAAUGGAACCAAAUCGACUGUCCAAUGGAUUUGGUCCCCUUCAACCAGCUGUAGCUGUGCAUUGAUGGCGCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002243
          miRBase ID: ptr-miR-133a
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-133c
          Stem-loop Accession # MI0014825
          Stem-loop Sequence
          GGGAGCCAAAUGCUUUGCUAGAGCUGGUAAAAUGGAACCAAAUCGACUGUCCAAUGGAUUUGGUCCCCUUCAACCAGCUGUAGCUGUGCAUUGAUGGCGCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0016973
          miRBase ID: ppy-miR-133c
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-133c
          Stem-loop Accession # MI0007621
          Stem-loop Sequence
          GGGAGCCAAAUGCUUUGCUAGAGCUGGUAAAAUGGAACCAAAUCGACUGUCCAAUGGAUUUGGUCCCCUUCAACCAGCUGUAGCUGUGCAUUGAUGGCGCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006777
          miRBase ID: mml-miR-133c-3p
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUG
          Chromosome Location: NA

          Tetraodon nigroviridis

          Stem-loop ID tni-mir-133
          Stem-loop Accession # MI0003398
          Stem-loop Sequence
          UGACUCCACAAUGCUUUGCUAAAGCUGGUAAAAUGGAACCAAAUCACCUCUUGAAUGGAUUUGGUCCCCUUCAACCAGCUGUAGCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003058
          miRBase ID: tni-miR-133
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUG
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-133a-1
          Stem-loop Accession # MI0001993
          Stem-loop Sequence
          CAUCAAACCACAAUGCUUUGCUAAAGCUGGUAAAAUGGAACCAAAUCACCUCUUCAAUGGAUUUGGUCCCCUUCAACCAGCUGUAGCUAUGCUUUGAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001830
          miRBase ID: dre-miR-133a-3p
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUG
          Chromosome Location: NA

          Stem-loop ID dre-mir-133a-2
          Stem-loop Accession # MI0001992
          Stem-loop Sequence
          AAUGCUUUGCUAAAGCUGGUAAAAUGGAACCAAAUCAACUGUUUUAUGGAUUUGGUCCCCUUCAACCAGCUGUAGCUGUGCAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001830
          miRBase ID: dre-miR-133a-3p
          Mature miRNA Sequence: UUUGGUCCCCUUCAACCAGCUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303121_pri
          TaqMan™ Pri-miRNA Assay : Hs03303117_pri
          TaqMan™ Pri-miRNA Assay : Mm03307401_pri
          TaqMan™ Pri-miRNA Assay : Rn03465790_pri
          TaqMan™ Pri-miRNA Assay : Mm03306281_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10413
          Ambion® Pre-miR™ miRNA Precursor : PM10413
          TaqMan™ MicroRNA Assay : 002246
          mirVana® miRNA inhibitor : MH10413
          mirVana® miRNA mimic : MC10413
          TaqMan™ Advanced miRNA Assay : 478511_mir
          TaqMan™ Advanced miRNA Assay : mmu481491_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-74676f44b6-nqctq:80/100.66.72.150:80.
          git-commit: db542085ed6beb0c3f0dd774c2548ab49b9f1b6a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026.10.42-1.0