Search
Search

MEP1A| Gene Symbol: MEP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005588.2 | NP_005579.2 | 7 | 452 | ||
| XM_011514628.1 | XP_011512930.1 | 6 | 509 | |||
| XM_011514629.1 | XP_011512931.1 | 7 | 452 | |||
| GenBank mRNA | AF478685.1 | - | 7 | 420 | ||
| AK225212.1 | - | 7 | 442 | |||
| AK290282.1 | - | 7 | 420 | |||
| AK315246.1 | - | 7 | 424 | |||
| AK315840.1 | - | 7 | 422 | |||
| BC131714.1 | - | 7 | 423 | 40122874 | ||
| BC143650.1 | - | 5 | 591 | |||
| BC143651.1 | - | 6 | 502 | 9052165 | ||
| DA432559.1 | - | 7 | 452 | |||
| M82962.1 | - | 7 | 419 | |||