Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DLG2 SIRNA Assays ›
Gene Symbol:
DLG2
Gene Name:
discs large MAGUK scaffold protein 2
Gene Aliases:
PPP1R58, PSD-93, PSD93, chapsyn-110
Chromosome Location:
Chr.11: 83455009 - 85628534 on Build GRCh38
UniGene ID:
Hs.367656
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
124294
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DLG2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142699.1
NP_001136171.1
28
7354
NM_001142700.1
NP_001136172.1
20
6389
NM_001142702.1
NP_001136174.1
11
5431
NM_001206769.1
NP_001193698.1
22
7038
NM_001300983.1
NP_001287912.1
23
6976
NM_001364.3
NP_001355.2
23
7030
XM_005273810.4
XP_005273867.1
22
6673
XM_005273811.4
XP_005273868.1
22
6619
XM_011544778.2
XP_011543080.1
29
8645
XM_011544780.2
XP_011543082.1
29
8591
XM_011544782.2
XP_011543084.1
24
7116
XM_011544788.2
XP_011543090.1
21
6463
XM_017017254.1
XP_016872743.1
30
8687
XM_017017255.1
XP_016872744.1
29
8648
XM_017017256.1
XP_016872745.1
29
8177
XM_017017257.1
XP_016872746.1
28
9109
XM_017017258.1
XP_016872747.1
27
8285
XM_017017259.1
XP_016872748.1
27
8006
XM_017017260.1
XP_016872749.1
26
8938
XM_017017261.1
XP_016872750.1
28
8435
XM_017017262.1
XP_016872751.1
23
7140
XM_017017263.1
XP_016872752.1
25
7158
XM_017017264.1
XP_016872753.1
24
7335
XM_017017265.1
XP_016872754.1
24
7332
XM_017017266.1
XP_016872755.1
22
7042
XM_017017267.1
XP_016872756.1
24
7240
XM_017017268.1
XP_016872757.1
24
7061
XM_017017269.1
XP_016872758.1
24
7762
XM_017017270.1
XP_016872759.1
24
7062
XM_017017271.1
XP_016872760.1
23
7239
XM_017017272.1
XP_016872761.1
23
7236
XM_017017273.1
XP_016872762.1
22
6991
XM_017017274.1
XP_016872763.1
24
7186
XM_017017275.1
XP_016872764.1
21
6941
XM_017017276.1
XP_016872765.1
23
7664
XM_017017277.1
XP_016872766.1
23
6984
XM_017017278.1
XP_016872767.1
21
6888
XM_017017279.1
XP_016872768.1
23
6906
XM_017017280.1
XP_016872769.1
23
7583
XM_017017281.1
XP_016872770.1
22
7083
XM_017017283.1
XP_016872772.1
22
6888
XM_017017284.1
XP_016872773.1
22
6560
XM_017017285.1
XP_016872774.1
22
7510
XM_017017286.1
XP_016872775.1
20
6424
XM_017017287.1
XP_016872776.1
13
5444
XM_017017288.1
XP_016872777.1
12
5283
XM_017017289.1
XP_016872778.1
12
6858
XM_017017290.1
XP_016872779.1
12
5503
XM_017017291.1
XP_016872780.1
11
5250
XM_017017292.1
XP_016872781.1
11
6762
GenBank mRNA
AK094585.1
-
1
1452
AK125151.1
-
1
2461
AK125152.1
-
1
1984
BC110844.1
-
1
1804
BX537627.1
-
1
1340
BX648386.1
-
1
2858
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
chemical synaptic transmission
nervous system development
negative regulation of phosphatase activity
sensory perception of pain
receptor clustering
establishment or maintenance of epithelial cell apical/basal polarity
GMP metabolic process
GDP metabolic process
receptor localization to synapse
guanylate kinase activity
protein binding
kinase binding
ionotropic glutamate receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-4sktr:80/100.66.76.202:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0