Search
Search

PLCB1| Gene Symbol: PLCB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015192.3 | NP_056007.1 | 6 | 894 | ||
| NM_182734.2 | NP_877398.1 | 6 | 894 | |||
| XM_011529199.2 | XP_011527501.1 | 6 | 854 | |||
| XM_011529200.2 | XP_011527502.1 | 4 | 518 | |||
| XM_011529201.2 | XP_011527503.1 | 4 | 328 | |||
| XM_017027752.1 | XP_016883241.1 | 6 | 854 | |||
| XM_017027753.1 | XP_016883242.1 | 6 | 481 | |||
| GenBank mRNA | AB011153.3 | - | 6 | 894 | ||
| AJ278313.1 | - | 6 | 510 | |||
| AJ278314.1 | - | 6 | 510 | |||
| AK127693.1 | - | 4 | 309 | |||
| AK299195.1 | - | 6 | 529 | |||
| AK310597.1 | - | 7 | 822 | |||
| AY004175.1 | - | 6 | 492 | |||
| BC069420.1 | - | 6 | 510 | 7262405 | ||
| BC117231.1 | - | 6 | 565 | 40125782 | ||