Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NRXN3 SIRNA Assays ›
Gene Symbol:
NRXN3
Gene Name:
neurexin 3
Gene Aliases:
C14orf60
Chromosome Location:
Chr.14: 77979904 - 79868290 on Build GRCh38
UniGene ID:
Hs.368307
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
137727
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NRXN3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001105250.2
NP_001098720.1
7
3010
NM_001272020.1
NP_001258949.1
6
3544
NM_004796.5
NP_004787.2
17
4373
NM_138970.4
NP_620426.2
6
2929
NR_073546.1
8
4322
NR_073547.1
21
7356
XM_005268218.2
XP_005268275.1
23
6737
XM_006720322.2
XP_006720385.1
22
6734
XM_006720323.2
XP_006720386.1
21
6398
XM_011537363.1
XP_011535665.1
23
6746
XM_011537364.2
XP_011535666.1
24
6708
XM_011537365.2
XP_011535667.1
23
6534
XM_011537366.1
XP_011535668.1
22
6722
XM_011537367.1
XP_011535669.1
22
6656
XM_011537368.1
XP_011535670.1
23
6443
XM_011537369.1
XP_011535671.1
23
6434
XM_011537370.1
XP_011535672.1
22
6410
XM_011537371.1
XP_011535673.1
23
6122
XM_011537372.1
XP_011535674.1
23
6113
XM_011537373.1
XP_011535675.1
22
6023
XM_011537377.2
XP_011535679.1
19
5038
XM_017021790.1
XP_016877279.1
24
6846
XM_017021791.1
XP_016877280.1
24
6629
XM_017021792.1
XP_016877281.1
22
6529
XM_017021793.1
XP_016877282.1
22
6719
XM_017021794.1
XP_016877283.1
21
6710
XM_017021795.1
XP_016877284.1
21
6505
XM_017021796.1
XP_016877285.1
21
6686
XM_017021797.1
XP_016877286.1
20
6686
XM_017021798.1
XP_016877287.1
20
6683
XM_017021799.1
XP_016877288.1
22
6647
XM_017021800.1
XP_016877289.1
22
6344
XM_017021801.1
XP_016877290.1
21
6317
XM_017021802.1
XP_016877291.1
20
6338
XM_017021803.1
XP_016877292.1
22
6092
XM_017021804.1
XP_016877293.1
22
6086
XM_017021805.1
XP_016877294.1
21
6077
XM_017021806.1
XP_016877295.1
21
6071
XM_017021807.1
XP_016877296.1
22
6032
GenBank mRNA
AL833739.1
-
6
2224
BC142649.1
-
7
2443
BC150194.1
-
7
2443
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neuron cell-cell adhesion
neurotransmitter secretion
axon guidance
synapse assembly
learning
adult behavior
social behavior
positive regulation of synapse assembly
vocalization behavior
positive regulation of synapse maturation
receptor activity
calcium channel regulator activity
metal ion binding
cell adhesion molecule binding
neuroligin family protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0