Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EPB41L1 SIRNA Assays ›
          Gene Symbol:
          EPB41L1
          Gene Name:
          erythrocyte membrane protein band 4.1 like 1
          Gene Aliases:
          4.1N, MRD11
          Chromosome Location:
          Chr.20: 36064891 - 36232799 on Build GRCh38
          UniGene ID:
          Hs.437422
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 146710

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: EPB41L1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001258329.1 NP_001245258.1 23 6134
          NM_001258330.1 NP_001245259.1 21 5552
          NM_001258331.1 NP_001245260.1 20 5779
          NM_012156.2 NP_036288.2 22 6083
          NM_177996.2 NP_818932.1 20 5774
          XM_011528667.1 XP_011526969.1 29 9066
          XM_011528671.1 XP_011526973.1 25 8153
          XM_011528673.1 XP_011526975.1 24 8113
          XM_011528677.1 XP_011526979.1 24 7927
          XM_011528681.1 XP_011526983.1 20 5887
          XM_011528682.1 XP_011526984.1 20 5884
          XM_011528683.1 XP_011526985.1 20 5842
          XM_011528685.1 XP_011526987.1 20 5515
          XM_017027711.1 XP_016883200.1 23 7876
          XM_017027712.1 XP_016883201.1 22 5977
          XM_017027713.1 XP_016883202.1 21 5920
          XM_017027714.1 XP_016883203.1 20 5833
          XM_017027715.1 XP_016883204.1 20 5887
          XM_017027716.1 XP_016883205.1 23 6113
          XM_017027717.1 XP_016883206.1 22 6071
          XM_017027718.1 XP_016883207.1 21 5542
          XM_017027719.1 XP_016883208.1 20 5481
          XM_017027720.1 XP_016883209.1 21 5656
          GenBank mRNA AB002336.1 - 22 6071
          BX537978.1 - 12 6499
          CR936825.1 - 19 5390

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          cortical actin cytoskeleton organization
          cell-cell adhesion
          actin binding
          structural molecule activity
          protein binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-bznxf:80/100.66.77.153:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline