Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Snupn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178374.3 | NP_848461.1 | 7 | 698 | ||
GenBank mRNA | AK002717.1 | - | 7 | 714 | ||
AK013151.1 | - | 7 | 708 | |||
AK155315.1 | - | 7 | 715 | |||
BC023317.1 | - | 678 | ||||
BC049638.1 | - | 7 | 698 | 6589249 | ||
BC061258.1 | - | 7 | 703 | 30251661 |