Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SLC9A3R1 SIRNA Assays ›
          Gene Symbol:
          Slc9a3r1
          Gene Name:
          solute carrier family 9 (sodium/hydrogen exchanger), member 3 regulator 1
          Gene Aliases:
          EBP-50, NHE-RF, NHERF-1, NHERF1
          Chromosome Location:
          Chr.11: 115163312 - 115181183 on Build GRCm38
          UniGene ID:
          Mm.27842
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 188258

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Slc9a3r1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012030.2 NP_036160.1 6 1336
          GenBank mRNA AK040371.1 - 200
          AK085210.1 - 6 1340
          AK153558.1 - 6 1336
          AK168894.1 - 6 1337
          BC085141.1 - 6 1321 6413140

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_021594, AF154336, FQ213792, FQ228926.

          Gene Ontology Categories:

          Function(s) Process(es)

          renal sodium ion transport
          adenylate cyclase-activating dopamine receptor signaling pathway
          sensory perception of sound
          negative regulation of cell proliferation
          negative regulation of platelet-derived growth factor receptor signaling pathway
          negative regulation of sodium ion transport
          negative regulation of phosphatidylinositol 3-kinase signaling
          Wnt signaling pathway
          cAMP-mediated signaling
          microvillus assembly
          actin cytoskeleton organization
          negative regulation of cell migration
          cellular phosphate ion homeostasis
          negative regulation of sodium:proton antiporter activity
          bile acid secretion
          cellular protein localization
          glutathione transport
          regulation of excretion
          regulation of protein kinase activity
          negative regulation of protein kinase B signaling
          auditory receptor cell stereocilium organization
          phospholipase C-activating dopamine receptor signaling pathway
          renal absorption
          negative regulation of ERK1 and ERK2 cascade
          renal phosphate ion absorption
          negative regulation of cell motility
          positive regulation of intrinsic apoptotic signaling pathway
          receptor binding
          protein binding
          beta-catenin binding
          chloride channel regulator activity
          phosphatase binding
          protein domain specific binding
          PDZ domain binding
          beta-2 adrenergic receptor binding
          protein complex binding
          protein complex scaffold
          protein self-association
          dopamine receptor binding
          growth factor receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0