Search
Search

Abcc1
LOC100912948| Gene Symbol: Abcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022281.2 | NP_071617.2 | 7 | 836 | ||
| GenBank mRNA | AF487549.1 | - | 7 | 836 | ||
| AY170916.1 | - | 7 | 797 | |||
| AY174892.1 | - | 7 | 836 | |||
| AY174893.1 | - | 7 | 836 | |||
| Gene Symbol: LOC100912948 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006245698.2 | XP_006245760.1 | 2 | 550 | ||