Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMCC1 SIRNA Assays ›
Gene Symbol:
TMCC1
Gene Name:
transmembrane and coiled-coil domain family 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 129647792 - 129893605 on Build GRCh38
UniGene ID:
Hs.477547
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
241573
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
286.00
Your Price
Online offer:
277.65
277.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMCC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001017395.3
NP_001017395.2
6
3187
NM_001128224.2
NP_001121696.1
6
2588
NR_033361.1
4
2733
XM_006713542.3
XP_006713605.1
7
3300
XM_006713543.3
XP_006713606.1
7
3297
XM_006713544.1
XP_006713607.1
7
3425
XM_006713550.3
XP_006713613.1
7
2858
XM_006713552.3
XP_006713615.1
4
2480
XM_006713553.3
XP_006713616.1
4
3772
XM_011512572.2
XP_011510874.1
8
3460
XM_011512573.2
XP_011510875.1
7
4039
XM_011512576.2
XP_011510878.1
7
3293
XM_011512579.2
XP_011510881.1
6
3048
XM_011512580.2
XP_011510882.1
9
3185
XM_011512581.2
XP_011510883.1
8
3022
XM_011512582.1
XP_011510884.1
4
2814
XM_011512583.2
XP_011510885.1
6
2591
XM_011512584.2
XP_011510886.1
8
2910
XM_017005931.1
XP_016861420.1
7
3208
XM_017005932.1
XP_016861421.1
8
3447
XM_017005933.1
XP_016861422.1
6
3244
XM_017005934.1
XP_016861423.1
10
3258
XM_017005935.1
XP_016861424.1
11
3347
XM_017005936.1
XP_016861425.1
8
3021
XM_017005937.1
XP_016861426.1
8
2955
XM_017005938.1
XP_016861427.1
10
3321
XM_017005939.1
XP_016861428.1
9
3215
XM_017005940.1
XP_016861429.1
6
2593
XM_017005941.1
XP_016861430.1
6
2719
GenBank mRNA
AB018322.1
-
3
1754
BC012480.1
-
3
1317
BC034284.1
-
3
1337
BC039859.1
-
6
3094
5527623
BX640712.1
-
4
2242
CR749206.1
-
4
2714
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_177412, AK173022, AK154540, AK036701.
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0