Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBRSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142641.1 | NP_001136113.1 | 17 | 3726 | ||
XM_005266171.3 | XP_005266228.1 | 20 | 3957 | |||
XM_005266173.3 | XP_005266230.1 | 20 | 3952 | |||
XM_005266175.3 | XP_005266232.1 | 19 | 3891 | |||
XM_005266176.3 | XP_005266233.1 | 19 | 3833 | |||
XM_005266177.3 | XP_005266234.1 | 20 | 3826 | |||
XM_011534803.2 | XP_011533105.1 | 20 | 4016 | |||
XM_011534804.2 | XP_011533106.1 | 20 | 4013 | |||
XM_011534805.2 | XP_011533107.1 | 19 | 3947 | |||
XM_011534806.2 | XP_011533108.1 | 19 | 3950 | |||
XM_011534807.2 | XP_011533109.1 | 19 | 3892 | |||
XM_011534808.2 | XP_011533110.1 | 20 | 3885 | |||
XM_011534809.2 | XP_011533111.1 | 19 | 3828 | |||
XM_011534810.2 | XP_011533112.1 | 19 | 3806 | |||
XM_011534812.2 | XP_011533114.1 | 21 | 3838 | |||
GenBank mRNA | AB046765.2 | - | 17 | 3497 |