Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300913.1 | NP_001287842.1 | 5 | 750 | ||
NM_022338.3 | NP_071733.1 | 4 | 1755 | |||
XM_005274053.3 | XP_005274110.1 | 4 | 1930 | |||
XM_011545103.2 | XP_011543405.1 | 2 | 1350 | |||
GenBank mRNA | AF264781.1 | - | 4 | 1715 | ||
AF370372.1 | - | 3 | 1623 | |||
AY358754.1 | - | 2 | 1317 | |||
BC011765.2 | - | 3 | 1604 | 3614861 | ||
BX384793.2 | - | 4 | 552 |