Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ALDOA SIRNA Assays ›
          Gene Symbol:
          Aldoa
          Gene Name:
          aldolase A, fructose-bisphosphate
          Gene Aliases:
          Aldo1, RNALDOG5
          Chromosome Location:
          Chr.1: 198228387 - 198233754 on Build Rnor_6.0
          UniGene ID:
          Rn.1774
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 47765

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Aldoa
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001177305.1 NP_001170776.1 3, 4 394
          NM_001271536.1 NP_001258465.1 3, 4 480
          NM_012495.2 NP_036627.1 3, 4 496
          XM_006230209.2 XP_006230271.1 5, 6 742
          XM_006230210.2 XP_006230272.1 4, 5 652
          XM_006230211.1 XP_006230273.1 4, 5 507
          GenBank mRNA BC064440.1 - 3, 4 473 5623628
          CF975930.1 - 3, 4 454
          CO398643.1 - 3, 4 387
          FQ212782.1 - 3, 4 447
          FQ213869.1 - 3, 4 477
          FQ214123.1 - 3, 4 447
          FQ214510.1 - 3, 4 449
          FQ216189.1 - 3, 4 402
          FQ216820.1 - 3, 4 398
          FQ224221.1 - 3, 4 487
          FQ228954.1 - 3, 4 426
          FQ229100.1 - 3, 4 441
          M12919.1 - 3, 4 496
          M14420.1 - 3, 4 394

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          aldolase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          fructose metabolic process
          glycolytic process
          ATP biosynthetic process
          striated muscle contraction
          regulation of cell shape
          response to heat
          fructose 1,6-bisphosphate metabolic process
          response to lipopolysaccharide
          response to nicotine
          response to estrogen
          muscle cell cellular homeostasis
          protein homotetramerization
          protease binding
          molecular_function
          fructose-bisphosphate aldolase activity
          protein binding
          cytoskeletal protein binding
          poly(A) RNA binding
          fructose binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-trq29:80/100.66.77.221:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0