Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145741.2 | NP_665684.2 | 2 | 917 | ||
GenBank mRNA | AK042684.1 | - | 908 | |||
BC022669.1 | - | 1 | 227 | |||
BC058358.1 | - | 2 | 917 | 6828166 | ||
L42114.1 | - | 2 | 733 | |||
S82648.1 | - | 2 | 733 |