Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NOX4 SIRNA Assays ›
          Gene Symbol:
          NOX4
          Gene Name:
          NADPH oxidase 4
          Gene Aliases:
          KOX, KOX-1, RENOX
          Chromosome Location:
          Chr.11: 89324353 - 89589611 on Build GRCh38
          UniGene ID:
          Hs.371036
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS121314
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NOX4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001143836.2 NP_001137308.1 7 845
          NM_001143837.1 NP_001137309.1 10 990
          NM_001291926.1 NP_001278855.1 7 841
          NM_001291927.1 NP_001278856.1 7 595
          NM_001291929.1 NP_001278858.1 6 508
          NM_001300995.1 NP_001287924.1 7 568
          NM_016931.4 NP_058627.1 7 845
          XM_006718849.3 XP_006718912.1 7 773
          XM_011542857.2 XP_011541159.1 7 645
          XM_017017841.1 XP_016873330.1 7 773
          XM_017017843.1 XP_016873332.1 7 773
          XM_017017844.1 XP_016873333.1 8 831
          XM_017017845.1 XP_016873334.1 4 247
          GenBank mRNA AB041035.1 - 7 765
          AF254621.1 - 7 612
          AF261943.1 - 7 526
          AJ704725.1 - 7 522
          AJ704726.1 - 7 526
          AJ704727.1 - 7 526
          AK057189.1 - 10 990
          AK291830.1 - 7 608
          AK298323.1 - 7 539
          AK298328.1 - 7 595
          AK298357.1 - 6 499
          AK298376.1 - 3 286
          AK311381.1 - 6 508
          AK311389.1 - 8 667
          AY288918.1 - 7 526
          BC040105.1 - 7 598 5590269
          BP376400.1 - 6 508

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          oxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to reactive oxygen species
          cell morphogenesis
          response to hypoxia
          superoxide metabolic process
          inflammatory response
          cell aging
          negative regulation of cell proliferation
          gene expression
          positive regulation of smooth muscle cell migration
          superoxide anion generation
          positive regulation of apoptotic process
          positive regulation of MAP kinase activity
          bone resorption
          homocysteine metabolic process
          positive regulation of stress fiber assembly
          positive regulation of protein kinase B signaling
          cardiac muscle cell differentiation
          oxidation-reduction process
          positive regulation of ERK1 and ERK2 cascade
          cellular response to cAMP
          cellular response to glucose stimulus
          cellular response to gamma radiation
          cellular response to transforming growth factor beta stimulus
          reactive oxygen species metabolic process
          positive regulation of reactive oxygen species metabolic process
          positive regulation of DNA biosynthetic process
          nucleotide binding
          protein binding
          electron carrier activity
          NAD(P)H oxidase activity
          superoxide-generating NADPH oxidase activity
          oxygen sensor activity
          heme binding
          flavin adenine dinucleotide binding
          modified amino acid binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0