Search
Search

RPUSD3| Gene Symbol: RPUSD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142547.1 | NP_001136019.1 | 4, 5 | 478 | ||
| NM_173659.3 | NP_775930.2 | 5, 6 | 523 | |||
| XM_011533627.2 | XP_011531929.1 | 5, 6 | 521 | |||
| GenBank mRNA | AK093620.1 | - | 2, 3 | 1701 | ||
| AK291888.1 | - | 5, 6 | 524 | |||
| AK296202.1 | - | 5, 6 | 510 | |||
| BC032135.2 | - | 5, 6 | 497 | 4649090 | ||
| BC062362.1 | - | 4, 5 | 454 | 5454417 | ||
| BC065741.1 | - | 5, 6 | 525 | 4091801 | ||