Search
Search

CD8B| Gene Symbol: CD8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004931.4 | NP_004922.1 | 3, 4 | 547 | ||
| NM_172101.3 | NP_742099.1 | 3, 4 | 547 | |||
| NM_172213.3 | NP_757362.1 | 3, 4 | 547 | |||
| XM_011533164.2 | XP_011531466.1 | 3, 4 | 505 | |||
| GenBank mRNA | AK303360.1 | - | 3, 4 | 505 | ||
| AK303595.1 | - | 2, 3 | 401 | |||
| BC100911.1 | - | 3, 4 | 538 | 40003720 | ||
| BC100913.1 | - | 3, 4 | 538 | 40003724 | ||
| BC100914.1 | - | 3, 4 | 538 | 40003725 | ||
| BQ004743.1 | - | 3, 4 | 494 | |||
| DC405359.1 | - | 3, 4 | 547 | |||
| M36712.1 | - | 3, 4 | 533 | |||
| X13444.1 | - | 3, 4 | 538 | |||
| X13445.1 | - | 3, 4 | 538 | |||
| X13452.1 | - | 3, 4 | 502 | |||
| Y00805.1 | - | 3, 4 | 533 | |||