Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZNRD1ASP SIRNA Assays ›
          Gene Symbol:
          ZNRD1ASP
          Gene Name:
          zinc ribbon domain containing 1 antisense, pseudogene
          Gene Aliases:
          C6orf12, HTEX4, NCRNA00171, TCTEX4, ZNRD1-AS, ZNRD1-AS1, ZNRD1AS, ZNRD1AS1
          Chromosome Location:
          Chr.6: 30001011 - 30061184 on Build GRCh38
          UniGene ID:
          Hs.653168
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n260731

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZNRD1ASP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_026751.1 3, 4 439
          GenBank mRNA AF238323.1 - 2, 3 977
          BC067890.1 - 3, 4 439
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BI462223.1 - 3, 4 445 Chr. 6 - 30002658 - 30061184 [-] on Build GRCh38
          GenBankmRNA DB079640.1 - 2, 3 261 Chr. 6 - 30021723 - 30061144 [-] on Build GRCh38
          GenBankmRNA DA107135.1 - 3, 4 412 Chr. 6 - 30035038 - 30061157 [-] on Build GRCh38
          GenBankmRNA BQ423269.1 - 2, 3 433 Chr. 6 - 30021686 - 30061095 [-] on Build GRCh38
          GenBankmRNA DA517191.1 - 3, 4 371 Chr. 6 - 30035024 - 30061116 [-] on Build GRCh38
          GenBankmRNA DB096670.1 - 3, 4 399 Chr. 6 - 30035023 - 30061144 [-] on Build GRCh38
          GenBankmRNA DA411941.1 - 2, 3 523 Chr. 6 - 30035937 - 30061184 [-] on Build GRCh38
          GenBankmRNA DA099095.1 - 3, 4 433 Chr. 6 - 30035068 - 30061178 [-] on Build GRCh38
          GenBankmRNA DB030030.1 - 2, 3 123 Chr. 6 - 29834970 - 30060784 [-] on Build GRCh38
          GenBankmRNA DA255293.1 - 2, 3 495 Chr. 6 - 30035915 - 30061157 [-] on Build GRCh38
          GenBankmRNA DA015305.1 - 3, 4 443 Chr. 6 - 30035041 - 30061184 [-] on Build GRCh38
          GenBankmRNA DA329344.1 - 2, 3 178 Chr. 6 - 30009812 - 30060840 [-] on Build GRCh38
          GenBankmRNA DB453565.2 - 3, 4 370 Chr. 6 - 30023736 - 30061141 [-] on Build GRCh38
          GenBankmRNA HY018590.1 - 3, 4 436 Chr. 6 - 30035928 - 30061206 [-] on Build GRCh38
          GenBankmRNA BQ434771.1 - 2, 3 508 Chr. 6 - 30021686 - 30061169 [-] on Build GRCh38
          GenBankmRNA AL705833.1 - 2, 3 458 Chr. 6 - 30034935 - 30058747 [-] on Build GRCh38
          GenBankmRNA DB061771.1 - 3, 4 441 Chr. 6 - 30035039 - 30061184 [-] on Build GRCh38
          GenBankmRNA DA507524.1 - 2, 3 355 Chr. 6 - 30034963 - 30061017 [-] on Build GRCh38
          GenBankmRNA DA734967.1 - 412 Chr. - - [-] on Build
          GenBankmRNA BG421163.1 - 384 Chr. - - [-] on Build

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-sbzgd:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0