Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TMEM198B SIRNA Assays ›
          Gene Symbol:
          TMEM198B
          Gene Name:
          transmembrane protein 198B (pseudogene)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 55829608 - 55836246 on Build GRCh38
          UniGene ID:
          Hs.731828
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n276635

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TMEM198B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_036476.1 2 1208
          NR_036477.1 2 1208
          NR_036478.1 2 1208
          NR_036479.1 2 1208
          GenBank mRNA AK057179.1 - 1 36
          AK095340.1 - 2 1071
          BC072431.1 - 1 96
          BX648969.1 - 1 50
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BI836674.1 - 1 122 Chr. 12 - 55830828 - 55835234 [+] on Build GRCh38
          GenBankmRNA DB226876.1 - 1 54 Chr. 12 - 55830896 - 55831649 [+] on Build GRCh38
          GenBankmRNA BI754435.1 - 1 92 Chr. 12 - 55830858 - 55834867 [+] on Build GRCh38
          GenBankmRNA DA978219.1 - 1 42 Chr. 12 - 55830908 - 55833247 [+] on Build GRCh38
          GenBankmRNA BF968189.1 - 1 96 Chr. 12 - 55830854 - 55835740 [+] on Build GRCh38
          GenBankmRNA DB055547.1 - 1 66 Chr. 12 - 55830884 - 55833243 [+] on Build GRCh38
          GenBankmRNA DA255899.1 - 1 74 Chr. 12 - 55830876 - 55834892 [+] on Build GRCh38
          GenBankmRNA DB268252.1 - 1 69 Chr. 12 - 55830881 - 55834944 [+] on Build GRCh38
          GenBankmRNA DA771305.1 - 1 110 Chr. 12 - 55830841 - 55831573 [+] on Build GRCh38
          GenBankmRNA DB035816.1 - 1 49 Chr. 12 - 55830901 - 55835050 [+] on Build GRCh38
          GenBankmRNA BQ880489.1 - 1 20 Chr. 12 - 55830930 - 55835235 [+] on Build GRCh38
          GenBankmRNA DA565769.1 - 1 110 Chr. 12 - 55830841 - 55833521 [+] on Build GRCh38
          GenBankmRNA BM919219.1 - 1 76 Chr. 12 - 55830893 - 55835439 [+] on Build GRCh38
          GenBankmRNA BM543506.1 - 1 94 Chr. 12 - 55830859 - 55835199 [+] on Build GRCh38
          GenBankmRNA DA880002.1 - 1 111 Chr. 12 - 55830839 - 55960602 [+] on Build GRCh38
          GenBankmRNA DA259378.1 - 1 65 Chr. 12 - 55830885 - 55834940 [+] on Build GRCh38
          GenBankmRNA DA873888.1 - 1 36 Chr. 12 - 55830915 - 55833256 [+] on Build GRCh38
          GenBankmRNA DA592104.1 - 1 76 Chr. 12 - 55830874 - 55833219 [+] on Build GRCh38
          GenBankmRNA BI768537.1 - 1 93 Chr. 12 - 55830858 - 55835272 [+] on Build GRCh38
          GenBankmRNA BI753624.1 - 1 102 Chr. 12 - 55830848 - 55835196 [+] on Build GRCh38
          GenBankmRNA DA962573.1 - 1 36 Chr. 12 - 55830915 - 55833212 [+] on Build GRCh38
          GenBankmRNA BG483291.1 - 1 50 Chr. 12 - 55830901 - 55833471 [+] on Build GRCh38
          GenBankmRNA DA237861.1 - 1 85 Chr. 12 - 55830865 - 55831582 [+] on Build GRCh38
          GenBankmRNA CN273340.1 - 1 108 Chr. 12 - 55830848 - 55833358 [+] on Build GRCh38
          GenBankmRNA DA866465.1 - 1 43 Chr. 12 - 55830907 - 55834942 [+] on Build GRCh38
          GenBankmRNA HY065428.1 - 1 64 Chr. 12 - 55830886 - 55833524 [+] on Build GRCh38
          GenBankmRNA BI768213.1 - 1 70 Chr. 12 - 55830880 - 55835188 [+] on Build GRCh38
          GenBankmRNA DT215055.1 - 1 47 Chr. 12 - 55830902 - 55835082 [+] on Build GRCh38
          GenBankmRNA DB175427.1 - 1 97 Chr. 12 - 55830853 - 55833196 [+] on Build GRCh38
          GenBankmRNA CV806966.1 - 1 95 Chr. 12 - 55830855 - 55833432 [+] on Build GRCh38
          GenBankmRNA BJ991497.1 - 1 108 Chr. 12 - 55830843 - 55833206 [+] on Build GRCh38
          GenBankmRNA DA346477.1 - 1 109 Chr. 12 - 55830841 - 55833188 [+] on Build GRCh38
          GenBankmRNA CN412989.1 - 1 185 Chr. 12 - 55830765 - 55834358 [+] on Build GRCh38
          GenBankmRNA DB138247.1 - 1 108 Chr. 12 - 55830843 - 55833588 [+] on Build GRCh38
          GenBankmRNA DB028630.1 - 1 111 Chr. 12 - 55830839 - 55834917 [+] on Build GRCh38
          GenBankmRNA DA561263.1 - 1 34 Chr. 12 - 55830916 - 55834988 [+] on Build GRCh38
          GenBankmRNA DA987504.1 - 1 83 Chr. 12 - 55830867 - 55833527 [+] on Build GRCh38
          GenBankmRNA DA929307.1 - 1 45 Chr. 12 - 55830905 - 55833246 [+] on Build GRCh38
          GenBankmRNA BI767471.1 - 92 Chr. - - [-] on Build
          GenBankmRNA DB204359.1 - 1 109 Chr. 12 - 55830841 - 55833195 [+] on Build GRCh38
          GenBankmRNA HY093156.1 - 1 33 Chr. 12 - 55830919 - 55833541 [+] on Build GRCh38
          GenBankmRNA BX458911.2 - 1 91 Chr. 12 - 55830859 - 55833521 [+] on Build GRCh38
          GenBankmRNA DA571226.1 - 1 81 Chr. 12 - 55830869 - 55833398 [+] on Build GRCh38
          GenBankmRNA DB008724.1 - 1 109 Chr. 12 - 55830841 - 55833177 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0