Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HS3ST6 SIRNA Assays ›
          Gene Symbol:
          Hs3st6
          Gene Name:
          heparan sulfate (glucosamine) 3-O-sulfotransferase 6
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 24752950 - 24758690 on Build GRCm38
          UniGene ID:
          Mm.328848
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s116256

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hs3st6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001012402.1 NP_001012402.1 2 903
          GenBank mRNA AK146974.1 - 3 1042
          AY574375.1 - 2 903
          BC132520.1 - 2 890 40130797
          BC138814.1 - 2 890 8861836
          BC145424.1 - 2 860 9053982
          BC145425.1 - 3 982

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Process(es)

          N-acetylglucosamine 6-O-sulfotransferase activity
          N-acetylgalactosamine 4-O-sulfotransferase activity
          heparan sulfate 2-O-sulfotransferase activity
          sulfotransferase activity
          [heparan sulfate]-glucosamine 3-sulfotransferase 1 activity
          HNK-1 sulfotransferase activity
          transferase activity
          heparan sulfate 6-O-sulfotransferase activity
          trans-9R,10R-dihydrodiolphenanthrene sulfotransferase activity
          1-phenanthrol sulfotransferase activity
          3-phenanthrol sulfotransferase activity
          4-phenanthrol sulfotransferase activity
          trans-3,4-dihydrodiolphenanthrene sulfotransferase activity
          9-phenanthrol sulfotransferase activity
          2-phenanthrol sulfotransferase activity
          phenanthrol sulfotransferase activity
          1-hydroxypyrene sulfotransferase activity
          galactose 3-O-sulfotransferase activity
          proteoglycan sulfotransferase activity
          cholesterol sulfotransferase activity
          hydroxyjasmonate sulfotransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0