Search
Search

Cetn4| Gene Symbol: Cetn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145825.2 | NP_665824.1 | 4 | 436 | ||
| GenBank mRNA | AF362367.1 | - | 4 | 436 | ||
| AK078275.1 | - | 3 | 2351 | |||
| AK131808.1 | - | 4 | 425 | |||
| BC038154.1 | - | 3 | 327 | |||
| BC060991.1 | - | 4 | 388 | 6742836 | ||
| BC087905.1 | - | 4 | 414 | 6707456 | ||