Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANK2 SIRNA Assays ›
Gene Symbol:
ANK2
Gene Name:
ankyrin 2, neuronal
Gene Aliases:
ANK-2, LQT4, brank-2
Chromosome Location:
Chr.4: 112706083 - 113383740 on Build GRCh38
UniGene ID:
Hs.620557
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s1365
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANK2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001127493.1
NP_001120965.1
46
5649
NM_001148.4
NP_001139.3
45
11810
NM_020977.3
NP_066187.2
44
5555
XM_005262945.2
XP_005263002.1
44
11809
XM_017008067.1
XP_016863556.1
48
12860
XM_017008068.1
XP_016863557.1
49
13101
XM_017008069.1
XP_016863558.1
47
12835
XM_017008070.1
XP_016863559.1
48
13046
XM_017008071.1
XP_016863560.1
49
12029
XM_017008072.1
XP_016863561.1
48
12005
XM_017008073.1
XP_016863562.1
46
12001
XM_017008074.1
XP_016863563.1
48
11975
XM_017008075.1
XP_016863564.1
47
11939
XM_017008076.1
XP_016863565.1
45
11908
XM_017008077.1
XP_016863566.1
45
11816
XM_017008078.1
XP_016863567.1
44
5653
XM_017008079.1
XP_016863568.1
43
5476
XM_017008080.1
XP_016863569.1
44
5504
XM_017008081.1
XP_016863570.1
44
5647
XM_017008082.1
XP_016863571.1
45
5731
XM_017008083.1
XP_016863572.1
46
5887
XM_017008084.1
XP_016863573.1
46
5782
XM_017008085.1
XP_016863574.1
46
5743
XM_017008086.1
XP_016863575.1
44
5551
XM_017008087.1
XP_016863576.1
46
5830
XM_017008088.1
XP_016863577.1
48
5717
XM_017008089.1
XP_016863578.1
44
5797
XM_017008090.1
XP_016863579.1
45
5794
XM_017008091.1
XP_016863580.1
43
5491
XM_017008092.1
XP_016863581.1
46
5684
XM_017008093.1
XP_016863582.1
47
5681
XM_017008094.1
XP_016863583.1
45
5650
XM_017008095.1
XP_016863584.1
43
5468
XM_017008096.1
XP_016863585.1
47
5624
XM_017008097.1
XP_016863586.1
44
5429
XM_017008098.1
XP_016863587.1
45
5591
XM_017008099.1
XP_016863588.1
46
5588
XM_017008100.1
XP_016863589.1
43
5393
XM_017008101.1
XP_016863590.1
43
5554
XM_017008102.1
XP_016863591.1
42
5369
XM_017008104.1
XP_016863593.1
44
5492
XM_017008105.1
XP_016863594.1
43
5468
XM_017008106.1
XP_016863595.1
42
5455
XM_017008107.1
XP_016863596.1
44
5429
XM_017008108.1
XP_016863597.1
42
5353
GenBank mRNA
AK294720.1
-
23
3300
AK299893.1
-
17
2397
BC125235.1
-
13
1533
BC125236.1
-
13
1533
BX537758.1
-
46
5623
BX538132.1
-
18
2552
BX538239.1
-
9
3715
X56958.2
-
44
5470
Z26634.2
-
45
11710
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of heart rate
atrial septum development
cellular calcium ion homeostasis
ER to Golgi vesicle-mediated transport
cytoskeleton organization
positive regulation of gene expression
regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
regulation of cardiac muscle contraction by calcium ion signaling
paranodal junction assembly
regulation of protein stability
T-tubule organization
protein localization to organelle
protein localization to cell surface
cellular protein localization
protein localization to M-band
protein localization to T-tubule
positive regulation of potassium ion transport
protein stabilization
regulation of release of sequestered calcium ion into cytosol
response to methylmercury
regulation of calcium ion transport
positive regulation of calcium ion transport
regulation of cardiac muscle contraction
regulation of ventricular cardiac muscle cell membrane repolarization
sarcoplasmic reticulum calcium ion transport
protein localization to endoplasmic reticulum
protein localization to plasma membrane
protein targeting to plasma membrane
regulation of cardiac muscle cell contraction
ventricular cardiac muscle cell action potential
atrial cardiac muscle cell action potential
SA node cell action potential
membrane depolarization during SA node cell action potential
atrial cardiac muscle cell to AV node cell communication
SA node cell to atrial cardiac muscle cell communication
regulation of heart rate by cardiac conduction
regulation of SA node cell action potential
regulation of atrial cardiac muscle cell action potential
positive regulation of potassium ion transmembrane transporter activity
regulation of calcium ion transmembrane transporter activity
positive regulation of calcium ion transmembrane transporter activity
positive regulation of cation channel activity
structural constituent of cytoskeleton
protein binding
cytoskeletal adaptor activity
enzyme binding
protein kinase binding
spectrin binding
protein binding, bridging
ion channel binding
ATPase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0