Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UBE3A SIRNA Assays ›
Gene Symbol:
UBE3A
Gene Name:
ubiquitin protein ligase E3A
Gene Aliases:
ANCR, AS, E6-AP, EPVE6AP, HPVE6A
Chromosome Location:
Chr.15: 25337234 - 25439086 on Build GRCh38
UniGene ID:
Hs.598862
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s14606
Validated
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
7,898.00
Your Price
Online offer:
7898.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: UBE3A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000462.3
NP_000453.2
7, 8
2383
NM_130838.1
NP_570853.1
3, 4
1598
NM_130839.2
NP_570854.1
6, 7
2318
XM_005268267.4
XP_005268324.1
8, 9
2685
XM_005268268.4
XP_005268325.1
7, 8
2572
XM_005268269.4
XP_005268326.1
8, 9
2416
XM_005268270.4
XP_005268327.1
7, 8
2373
XM_005268271.4
XP_005268328.1
4, 5
2192
XM_006720675.3
XP_006720738.1
8, 9
2637
XM_006720676.3
XP_006720739.1
5, 6
1882
XM_011521994.2
XP_011520296.1
8, 9
2574
XM_011521995.2
XP_011520297.1
8, 9
2248
XM_017022544.1
XP_016878033.1
9, 10
2610
XM_017022545.1
XP_016878034.1
8, 9
2569
XM_017022546.1
XP_016878035.1
8, 9
2477
XM_017022547.1
XP_016878036.1
7, 8
2509
XM_017022548.1
XP_016878037.1
7, 8
2504
XM_017022549.1
XP_016878038.1
7, 8
2311
XM_017022550.1
XP_016878039.1
7, 8
2463
XM_017022551.1
XP_016878040.1
8, 9
2514
XM_017022552.1
XP_016878041.1
8, 9
2204
XM_017022553.1
XP_016878042.1
8, 9
2519
XM_017022554.1
XP_016878043.1
4, 5
2523
XM_017022555.1
XP_016878044.1
6, 7
2292
XM_017022556.1
XP_016878045.1
7, 8
2441
XM_017022557.1
XP_016878046.1
6, 7
2376
XM_017022558.1
XP_016878047.1
7, 8
2572
XM_017022559.1
XP_016878048.1
4, 5
2192
GenBank mRNA
AB194232.1
-
5, 6
1694
AF273050.1
-
4, 5
1653
AK130491.1
-
2, 3
1327
AK291405.1
-
4, 5
1744
AK292514.1
-
6, 7
1928
BC002582.2
-
6, 7
2271
3160225
BC009271.1
-
1, 2
875
3353595
L07557.1
-
3, 4
1620
U84404.1
-
8, 9
2140
X98031.1
-
4, 5
1623
X98032.1
-
2, 3
1554
X98033.1
-
4, 5
1614
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001191837, XM_008759399, XM_006229313, XM_006229310, XM_008759400, and others.
Matching Taqman Gene Expression Assay:
Hs00166580_m1
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
ovarian follicle development
proteolysis
ubiquitin-dependent protein catabolic process
brain development
positive regulation of phosphatidylinositol 3-kinase signaling
viral process
androgen receptor signaling pathway
positive regulation of protein ubiquitination
sperm entry
regulation of circadian rhythm
positive regulation of transcription from RNA polymerase II promoter
protein autoubiquitination
prostate gland growth
protein K48-linked ubiquitination
regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
transcription coactivator activity
ubiquitin-protein transferase activity
protein binding
ligase activity
metal ion binding
ubiquitin protein ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0