Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SORBS2 SIRNA Assays ›
Gene Symbol:
SORBS2
Gene Name:
sorbin and SH3 domain containing 2
Gene Aliases:
ARGBP2, PRO0618
Chromosome Location:
Chr.4: 185585444 - 185956716 on Build GRCh38
UniGene ID:
Hs.481342
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s16088
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SORBS2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145670.1
NP_001139142.1
13
1888
NM_001145671.2
NP_001139143.1
15
1677
NM_001145672.1
NP_001139144.1
13
1315
NM_001145673.1
NP_001139145.1
15
2201
NM_001145674.1
NP_001139146.1
6
907
NM_001145675.1
NP_001139147.1
7
952
NM_001270771.1
NP_001257700.1
15
1942
NM_003603.6
NP_003594.3
15
1916
NM_021069.4
NP_066547.1
12
1722
XM_005263307.1
XP_005263364.1
17
2795
XM_005263308.1
XP_005263365.1
16
2153
XM_005263310.1
XP_005263367.1
16
2009
XM_005263311.1
XP_005263368.1
15
1994
XM_005263312.1
XP_005263369.1
15
1991
XM_005263313.1
XP_005263370.1
13
1892
XM_006714388.1
XP_006714451.1
16
2084
XM_006714390.1
XP_006714453.1
14
1901
XM_017008738.1
XP_016864227.1
19
3353
XM_017008739.1
XP_016864228.1
20
3656
XM_017008740.1
XP_016864229.1
19
3349
XM_017008741.1
XP_016864230.1
19
3353
XM_017008742.1
XP_016864231.1
18
3303
XM_017008743.1
XP_016864232.1
19
3284
XM_017008744.1
XP_016864233.1
19
3563
XM_017008745.1
XP_016864234.1
18
3503
XM_017008746.1
XP_016864235.1
18
3194
XM_017008747.1
XP_016864236.1
19
3494
XM_017008748.1
XP_016864237.1
16
3053
XM_017008749.1
XP_016864238.1
19
2915
XM_017008750.1
XP_016864239.1
19
3353
XM_017008751.1
XP_016864240.1
18
3269
XM_017008752.1
XP_016864241.1
18
3194
XM_017008753.1
XP_016864242.1
16
3053
XM_017008754.1
XP_016864243.1
15
3008
XM_017008755.1
XP_016864244.1
13
2018
XM_017008756.1
XP_016864245.1
12
1721
XM_017008757.1
XP_016864246.1
8
1123
XM_017008758.1
XP_016864247.1
16
2187
XM_017008759.1
XP_016864248.1
13
2614
XM_017008760.1
XP_016864249.1
15
2060
XM_017008762.1
XP_016864251.1
14
1937
XM_017008763.1
XP_016864252.1
13
1887
XM_017008764.1
XP_016864253.1
14
1868
XM_017008765.1
XP_016864254.1
15
2171
XM_017008766.1
XP_016864255.1
13
1844
XM_017008767.1
XP_016864256.1
14
2147
XM_017008768.1
XP_016864257.1
13
2065
XM_017008769.1
XP_016864258.1
13
1823
XM_017008770.1
XP_016864259.1
12
1799
XM_017008771.1
XP_016864260.1
14
1941
XM_017008772.1
XP_016864261.1
13
1775
XM_017008773.1
XP_016864262.1
14
2078
XM_017008774.1
XP_016864263.1
9
1568
XM_017008775.1
XP_016864264.1
18
3308
GenBank mRNA
AB018320.1
-
12
1374
AF049884.1
-
15
1880
AF049885.1
-
14
1320
AK056628.1
-
15
1560
AK056758.1
-
8
1123
AK225327.1
-
13
1854
AK225812.1
-
15
2201
AK293400.1
-
13
1315
AK295742.1
-
13
1293
AK296461.1
-
6
684
AK304653.1
-
7
688
BC011883.1
-
15
1644
3677070
EF139852.1
-
1
20
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
actin filament organization
cell adhesion
biological_process
cell migration
cell growth involved in cardiac muscle cell development
structural constituent of cytoskeleton
protein binding
cytoskeletal adaptor activity
structural constituent of muscle
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0