Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other INHA SIRNA Assays ›
          Gene Symbol:
          INHA
          Gene Name:
          inhibin alpha subunit
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 219572232 - 219575713 on Build GRCh38
          UniGene ID:
          Hs.407506
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s194558

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: INHA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002191.3 NP_002182.1 2 1176
          GenBank mRNA BC006391.2 - 2 1113 4126990
          BT006954.1 - 2 1033
          M13144.1 - 2 990
          M13981.1 - 2 1177

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          ovarian follicle development
          cell cycle arrest
          signal transduction
          cell surface receptor signaling pathway
          cell-cell signaling
          nervous system development
          male gonad development
          response to external stimulus
          positive regulation of pathway-restricted SMAD protein phosphorylation
          cell differentiation
          erythrocyte differentiation
          regulation of cell proliferation
          negative regulation of phosphorylation
          hemoglobin biosynthetic process
          regulation of apoptotic process
          regulation of MAPK cascade
          negative regulation of interferon-gamma biosynthetic process
          negative regulation of B cell differentiation
          negative regulation of macrophage differentiation
          negative regulation of cell cycle
          positive regulation of follicle-stimulating hormone secretion
          negative regulation of follicle-stimulating hormone secretion
          cell development
          regulation of cell cycle
          SMAD protein signal transduction
          receptor binding
          cytokine activity
          transforming growth factor beta receptor binding
          hormone activity
          protein binding
          growth factor activity
          inhibin binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-z8nhc:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0