Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SULT1A1 SIRNA Assays ›
          Gene Symbol:
          SULT1A1 SLX1A-SULT1A3 SLX1B-SULT1A4 SULT1A2 SULT1A3 SULT1A4
          Gene Name:
          sulfotransferase family 1A member 1
          SLX1A-SULT1A3 readthrough (NMD candidate)
          SLX1B-SULT1A4 readthrough (NMD candidate)
          sulfotransferase family 1A member 2
          sulfotransferase family 1A member 3
          sulfotransferase family 1A member 4
          Gene Aliases:
          HAST1/HAST2, P-PST, PST, ST1A1, ST1A3, STP, STP1, TSPST1
          -
          -
          HAST4, P-PST, ST1A2, STP2, TSPST2
          HAST, HAST3, M-PST, ST1A3, ST1A3/ST1A4, ST1A5, STM, TL-PST
          HAST3, M-PST, ST1A3/ST1A4, ST1A4, TL-PST
          Chromosome Location:
          Chr.16: 28605587 - 28623586 on Build GRCh38
          Chromosome Location:
          Chr.16: 30194433 - 30204329 on Build GRCh38
          Chromosome Location:
          Chr.16: 29455091 - 29464980 on Build GRCh38
          Chromosome Location:
          Chr.16: 28591943 - 28597079 on Build GRCh38
          Chromosome Location:
          Chr.16: 30199228 - 30204329 on Build GRCh38
          Chromosome Location:
          Chr.16: 29459886 - 29464980 on Build GRCh38
          UniGene ID:
          Hs.567342
          Hs.460558
          Hs.744871
          Hs.546304
          Hs.460558
          Hs.460558
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          siRNA ID s194825

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SULT1A1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001055.3 NP_001046.2 2 189
          NM_177529.2 NP_803565.1 2 220
          NM_177530.2 NP_803566.1 2 345
          NM_177534.2 NP_803878.1 1 544
          XM_017023604.1 XP_016879093.1 3 274
          XM_017023605.1 XP_016879094.1 3 271
          XM_017023606.1 XP_016879095.1 3 274
          XM_017023607.1 XP_016879096.1 5 835
          XM_017023608.1 XP_016879097.1 2 189
          XM_017023609.1 XP_016879098.1 2 239
          XM_017023610.1 XP_016879099.1 2 270
          XM_017023611.1 XP_016879100.1 4 762
          XM_017023612.1 XP_016879101.1 5 686
          XM_017023613.1 XP_016879102.1 3 501
          GenBank mRNA AB062428.1 - 1 127
          AB209149.1 - 5497
          AJ007418.1 - 1 88
          BC000923.2 - 2 139 3447900
          BC110887.1 - 5 602 5190558
          BG482944.1 - 2 190
          BG990719.1 - 2 266
          BM927749.1 - 2 166
          BT007324.1 - 1 71
          CV023878.1 - 1 71
          L10819.1 - 2 109
          L19955.1 - 1 112
          L19999.1 - 2 141
          U09031.1 - 1 112
          U26309.1 - 1 136
          X78283.1 - 2 230
          X84654.1 - 1 85
          Gene Symbol: SLX1A-SULT1A3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_037608.1 5 992
          GenBank mRNA AK094769.1 - 3 1474
          AK097386.1 - 4 638
          AK122707.1 - 5 993
          AK122733.1 - 6 820
          BC014471.1 - 3 257 4907290
          BC111011.1 - 4 1020 5768190
          X84653.1 - 2 92
          Gene Symbol: SLX1B-SULT1A4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_037609.1 5 992
          Gene Symbol: SULT1A2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001054.3 NP_001045.1 2 149
          NM_177528.2 NP_803564.1 2 422
          XM_006721077.3 XP_006721140.1 1 785
          XM_017023600.1 XP_016879089.1 2 158
          XM_017023601.1 XP_016879090.1 2 586
          XM_017023602.1 XP_016879091.1 3 288
          XM_017023603.1 XP_016879092.1 2 154
          GenBank mRNA BC113727.1 - 2 144 8322779
          BC113729.1 - 2 144 8322781
          U28169.1 - 2 144
          U28170.1 - 2 132
          X78282.1 - 1 71
          Gene Symbol: SULT1A3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_177552.3 NP_808220.1 2 175
          GenBank mRNA AB111093.1 - 1 71
          AB111094.1 - 1 71
          AB111095.1 - 1 71
          AB111096.1 - 1 71
          AB111097.1 - 1 71
          AB111098.1 - 1 71
          AB111099.1 - 1 71
          AK298073.1 - 2 176
          BC078144.1 - 2 119 6022425
          BC132914.1 - 2 172 40146936
          BC132920.1 - 2 172 40146942
          BC144354.1 - 2 172 9052874
          BC144355.1 - 2 172 9052875
          BC171772.1 - 2 172 9054426
          DA978913.1 - 2 177
          L25275.1 - 2 135
          U08032.1 - 1 77
          U34199.1 - 1 71
          Gene Symbol: SULT1A4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001017390.2 NP_001017390.1 2 175

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          catecholamine metabolic process
          xenobiotic metabolic process
          steroid metabolic process
          estrogen metabolic process
          amine metabolic process
          flavonoid metabolic process
          3'-phosphoadenosine 5'-phosphosulfate metabolic process
          sulfation
          amine biosynthetic process
          phenol-containing compound metabolic process
          IRE1-mediated unfolded protein response
          aryl sulfotransferase activity
          protein binding
          sulfotransferase activity
          flavonol 3-sulfotransferase activity
          steroid sulfotransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0