Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ANKRD36 SIRNA Assays ›
          Gene Symbol:
          ANKRD36 ANKRD36B ANKRD36C
          Gene Name:
          ankyrin repeat domain 36
          ankyrin repeat domain 36B
          ankyrin repeat domain 36C
          Gene Aliases:
          UNQ2430
          KIAA1641
          -
          Chromosome Location:
          Chr.2: 97113186 - 97264521 on Build GRCh38
          Chromosome Location:
          Chr.2: 97492663 - 97590245 on Build GRCh38
          Chromosome Location:
          Chr.2: 95849548 - 95992320 on Build GRCh38
          UniGene ID:
          Hs.541894
          Hs.532921
          Hs.646318
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID s198481

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ANKRD36
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006712514.3 XP_006712577.1 1 220
          XM_011511130.1 XP_011509432.1 1 220
          XM_011511131.1 XP_011509433.1 1 220
          XM_017004009.1 XP_016859498.1 1 220
          XM_017004010.1 XP_016859499.1 1 220
          XM_017004011.1 XP_016859500.1 1 220
          XM_017004012.1 XP_016859501.1 1 220
          XM_017004013.1 XP_016859502.1 1 220
          XM_017004014.1 XP_016859503.1 1 220
          XM_017004015.1 XP_016859504.1 1 220
          XM_017004016.1 XP_016859505.1 1 220
          XM_017004017.1 XP_016859506.1 1 220
          XM_017004018.1 XP_016859507.1 1 220
          XM_017004019.1 XP_016859508.1 1 220
          XM_017004020.1 XP_016859509.1 1 220
          XM_017004021.1 XP_016859510.1 1 220
          XM_017004022.1 XP_016859511.1 1 220
          XM_017004023.1 XP_016859512.1 1 220
          XM_017004024.1 XP_016859513.1 1 220
          XM_017004025.1 XP_016859514.1 1 220
          XM_017004026.1 XP_016859515.1 1 220
          XM_017004027.1 XP_016859516.1 1 220
          XM_017004028.1 XP_016859517.1 1 220
          XM_017004029.1 XP_016859518.1 1 220
          XM_017004030.1 XP_016859519.1 1 220
          XM_017004031.1 XP_016859520.1 1 220
          XM_017004032.1 XP_016859521.1 1 220
          XM_017004033.1 XP_016859522.1 1 220
          XM_017004034.1 XP_016859523.1 1 220
          XM_017004035.1 XP_016859524.1 1 220
          XM_017004036.1 XP_016859525.1 1 220
          XM_017004037.1 XP_016859526.1 1 220
          XM_017004038.1 XP_016859527.1 1 220
          XM_017004039.1 XP_016859528.1 1 220
          XM_017004040.1 XP_016859529.1 1 220
          XM_017004041.1 XP_016859530.1 1 220
          XM_017004042.1 XP_016859531.1 1 220
          XM_017004043.1 XP_016859532.1 1 220
          XM_017004044.1 XP_016859533.1 1 220
          XM_017004045.1 XP_016859534.1 1 220
          XM_017004046.1 XP_016859535.1 1 220
          XM_017004047.1 XP_016859536.1 1 220
          XM_017004048.1 XP_016859537.1 1 220
          XM_017004049.1 XP_016859538.1 1 220
          Gene Symbol: ANKRD36B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_005263995.4 XP_005264052.1 1 190
          XM_006712661.1 XP_006712724.1 1 190
          XM_011511555.1 XP_011509857.1 1 190
          XM_011511556.1 XP_011509858.1 1 190
          XM_011511557.2 XP_011509859.1 1 190
          XM_011511558.1 XP_011509860.1 1 190
          XM_017004578.1 XP_016860067.1 1 190
          XM_017004579.1 XP_016860068.1 1 190
          XM_017004580.1 XP_016860069.1 1 190
          XM_017004581.1 XP_016860070.1 1 190
          XM_017004582.1 XP_016860071.1 1 190
          XM_017004583.1 XP_016860072.1 1 190
          XM_017004584.1 XP_016860073.1 1 190
          XM_017004585.1 XP_016860074.1 1 190
          XM_017004587.1 XP_016860076.1 1 190
          XM_017004588.1 XP_016860077.1 1 190
          XM_017004589.1 XP_016860078.1 1 190
          XM_017004590.1 XP_016860079.1 1 190
          XM_017004592.1 XP_016860081.1 1 190
          XM_017004593.1 XP_016860082.1 1 190
          XM_017004594.1 XP_016860083.1 1 190
          XM_017004595.1 XP_016860084.1 1 190
          Gene Symbol: ANKRD36C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001310154.1 NP_001297083.1 1 278

          Back To Top

          More Information


          Gene Ontology Categories:

          Process(es)

          protein binding
          ion channel inhibitor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0