Search
Search

CKS1B| Gene Symbol: CKS1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001826.2 | NP_001817.1 | 3 | 672 | ||
| NR_024163.1 | 3 | 780 | ||||
| GenBank mRNA | AF274943.1 | - | 3 | 600 | ||
| AF279897.1 | - | 3 | 595 | |||
| AK307066.1 | - | 3 | 728 | |||
| BC007751.2 | - | 3 | 615 | 4110344 | ||
| BC015629.2 | - | 3 | 594 | 3162564 | ||
| BC070319.1 | - | 3 | 633 | 4738841 | ||
| BC070320.1 | - | 3 | 633 | 6494754 | ||
| BQ883992.1 | - | 3 | 611 | |||
| X54941.1 | - | 3 | 577 | |||