Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018047.2 | NP_060517.1 | 8 | 906 | ||
GenBank mRNA | AK001152.1 | - | 8 | 861 | ||
AK096728.1 | - | 1 | 631 | |||
AK297019.1 | - | 7 | 675 | |||
AL136933.1 | - | 8 | 835 | |||
BC003402.1 | - | 8 | 839 | 3449258 |