Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EEF2 SIRNA Assays ›
          Gene Symbol:
          Eef2
          Gene Name:
          eukaryotic translation elongation factor 2
          Gene Aliases:
          Ef-2
          Chromosome Location:
          Chr.10: 81176631 - 81182509 on Build GRCm38
          UniGene ID:
          Mm.2903
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s65330

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Eef2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_007907.2 NP_031933.1 8 1403
          GenBank mRNA AK028938.1 - 8 1382
          AK032997.1 - 8 1389
          AK040474.1 - 8 1389
          AK077866.1 - 8 1389
          AK087985.1 - 8 1389
          AK128976.1 - 8 1388
          AK134195.1 - 8 1389
          AK144874.1 - 8 1391
          AK145509.1 - 8 1390
          AK145545.1 - 8 1389
          AK146508.1 - 8 1391
          AK146947.1 - 8 1389
          AK149659.1 - 8 1390
          AK150136.1 - 8 1389
          AK150902.1 - 8 1389
          AK151487.1 - 8 1389
          AK151812.1 - 8 1339
          AK151945.1 - 8 1389
          AK152146.1 - 8 1389
          AK152447.1 - 8 1389
          AK152587.1 - 8 1389
          AK152826.1 - 8 1389
          AK153275.1 - 8 1389
          AK153936.1 - 8 1389
          AK155325.1 - 8 1389
          AK159155.1 - 8 1390
          AK159421.1 - 8 1389
          AK159514.1 - 8 1389
          AK159806.1 - 8 1389
          AK159829.1 - 8 1389
          AK163063.1 - 8 1389
          AK166212.1 - 8 1389
          AK166595.1 - 8 1389
          AK166596.1 - 8 1389
          AK166719.1 - 8 1390
          AK166747.1 - 8 1389
          AK166751.1 - 8 1387
          AK166758.1 - 8 1390
          AK166788.1 - 8 1390
          AK166815.1 - 8 1390
          AK166829.1 - 8 1390
          AK166851.1 - 8 1387
          AK166968.1 - 8 1389
          AK167081.1 - 8 1390
          AK167093.1 - 8 1389
          AK167109.1 - 8 1389
          AK167115.1 - 8 1389
          AK167199.1 - 8 1389
          AK167208.1 - 8 1387
          AK167221.1 - 8 1389
          AK167230.1 - 8 1389
          AK167281.1 - 8 1389
          AK168527.1 - 8 1390
          AK168615.1 - 8 1390
          AK168638.1 - 8 1390
          AK168659.1 - 8 1390
          AK168794.1 - 8 1389
          AK168827.1 - 8 1389
          AK168874.1 - 8 1382
          AK169013.1 - 8 1389
          AK191254.1 - 1 26
          AK192929.1 - 26
          AK197735.1 - 2 329
          AK199168.1 - 1 26
          AK202379.1 - 1 82
          AK205796.1 - 2 284
          AK210206.1 - 1 26
          AK211322.1 - 1 83
          AK215832.1 - 2 284
          BC007152.1 - 8 1387 3600352
          BC013263.1 - 8 1375
          BC060707.1 - 7 1264
          U89416.1 - 1 24

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          translation elongation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          hematopoietic progenitor cell differentiation
          translation
          translational elongation
          positive regulation of translation
          nucleotide binding
          translation elongation factor activity
          GTPase activity
          protein binding
          GTP binding
          protein kinase binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0