Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CAD SIRNA Assays ›
          Gene Symbol:
          Cad
          Gene Name:
          carbamoyl-phosphate synthetase 2, aspartate transcarbamylase, and dihydroorotase
          Gene Aliases:
          2410008J01Rik, AU018859, Cpad
          Chromosome Location:
          Chr.5: 31054636 - 31078479 on Build GRCm38
          UniGene ID:
          Mm.305535
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s88143

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Cad
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_023525.2 NP_076014.1 39 6172
          GenBank mRNA AK010453.1 - 1 26
          BC026620.1 - 9 1101
          BC043325.1 - 12 1701
          BC060119.1 - 39 6138
          BC060717.1 - 30 4613
          BC137856.1 - 39 6024 8860875

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          hydrolase ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          urea cycle
          'de novo' pyrimidine nucleobase biosynthetic process
          pyrimidine nucleotide biosynthetic process
          UTP biosynthetic process
          cellular amino acid metabolic process
          arginine biosynthetic process
          glutamine metabolic process
          nitrogen compound metabolic process
          metabolic process
          drug metabolic process
          peptidyl-threonine phosphorylation
          protein autophosphorylation
          nucleotide binding
          UTP binding
          catalytic activity
          aspartate carbamoyltransferase activity
          carbamoyl-phosphate synthase (ammonia) activity
          carbamoyl-phosphate synthase (glutamine-hydrolyzing) activity
          dihydroorotase activity
          protein kinase activity
          ATP binding
          zinc ion binding
          amino acid binding
          transferase activity
          carboxyl- or carbamoyltransferase activity
          hydrolase activity
          hydrolase activity, acting on carbon-nitrogen (but not peptide) bonds
          hydrolase activity, acting on carbon-nitrogen (but not peptide) bonds, in cyclic amides
          ligase activity
          enzyme binding
          identical protein binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0