    • See all product categories
  • Applications
    • Cell Analysis
    • Protein Biology
    • Lab Informatics
    • 360° CDMO and CRO Solutions
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
            • Services Central
          • Proteins & Peptides ›
          • Eotaxin 2 Proteins
          Gibco
          Mouse CCL24 (Eotaxin-2) Recombinant Protein, PeproTech®
          9 References
          View all (3) Eotaxin 2 proteins
          Tech Support
          Cite Mouse CCL24 (Eotaxin-2) Recombinant Protein, PeproTech®
          Additional Information:
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Testing Data (1)
          Mouse CCL24 (Eotaxin-2) Protein in Bioactivity (Bioactivity)
          Group 53 Created with Sketch.
          Mouse CCL24 (Eotaxin-2) Protein in Bioactivity (Bioactivity)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Mouse CCL24 (Eotaxin-2) Protein (250-22-250UG) in Bioactivity
          Bioassay analysis of Mouse CCL24 (Eotaxin-2) Recombinant Protein, PeproTech® (Product # 250-22-1MG). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Mouse CCL24 (Eotaxin-2) Protein in Bioactivity (Bioactivity)
          Product Details
          250-22-250UG
          Applications
          Tested Dilution
          Publications
          Control (Ctrl)
          Assay-dependent
          -
          In vitro Assay (In vitro)
          -
          View 4 publications 4 publications
          Miscellaneous PubMed (Misc)
          -
          View 4 publications 4 publications
          Western blot control (WB Ctrl)
          Assay-dependent
          -
          Bioactivity (Bioactivity)
          Assay-dependent
          -
          Cell induction (Cell Ind)
          -
          View 1 publication 1 publication
          Product Specifications
          Species
          Mouse
          Published species
          Human, Mouse
          Expression System
          E. coli
          Amino acid sequence
          VTIPSSCCTS FISKKIPENR VVSYQLANGS ICPKAGVIFI TKKGHKICTD PKLLWVQRHI QKLDAKKNQP SKGAKAVRTK FAVQRRRGNS TEV
          Molecular weight
          10.3 kDa
          Class
          Recombinant
          Type
          Protein
          Purity
          ≥ 98% by SDS-PAGE gel and HPLC analyses.
          Endotoxin concentration
          <1 EU/µg
          Activity
          Determined by its ability to chemoattract murine lymphocytes using a concentration range of 10-100 ng/ml.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Lyophilized
          Amount
          250 µg
          Purification
          purified
          Contains
          no preservative
          Storage conditions
          -20°C
          Shipping conditions
          Ambient
          Product Specific Information
          Recombinant Murine Eotaxin-2 is a 10.3 kDa protein containing 93 amino acid residues.
          This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
          Target Information
          Eotaxin 2 belongs to the subfamily of small cytokine CC genes. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity on resting T lymphocytes, a minimal activity on neutrophils, and is negative on monocytes and activated T lymphocytes. The protein is also a strong suppressor of colony formation by a multipotential hematopoietic progenitor cell line.
          Bioinformatics
          Protein Aliases: CC chemokine; CC chemokine CCL24; chemokine (C-C motif) ligand 24; eosinophil chemotactic protein 2; eotaxin-2; Eotaxin2; small-inducible cytokine A24; unnamed protein product
          View more View less
          Gene Aliases: CKb-6; MPIF-2; Scya24
          View more View less
          UniProt ID: (Mouse) Q9JKC0
          View more View less
          Entrez Gene ID: (Mouse) 56221
          • Trademarks
          • Fair Trade Fair Trade
          Korea flag icon
          Korea
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910
          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783
          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)
          ISMS Logo
          Your items have has been added!
          Host server : magellan-search-green-79565875f6-l699q:80/100.66.79.160:80.
          git-commit: 280dcbe45276d5d6e613c14d399020b8a49b7116
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.57.0-Offline